Skip to Content
Merck
All Photos(1)

Documents

SAB4502013

Sigma-Aldrich

Anti-ENOS antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Constitutive NOS, EC-NOS, ECNOS, Endothelial NOS, cNOS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 133 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS3(4846)

General description

Anti-ENOS Antibody detects endogenous levels of total ENOS protein.
The ENOS (endothelial nitric oxide synthase) gene is mapped to human chromosome 7q36.1. ENOS is expressed predominantly in the endothelial cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human eNOS.

Immunogen Range: 581-630

Application

Anti-ENOS antibody produced in rabbit has been used in SDS-PAGE and western blotting.

Biochem/physiol Actions

ENOS (endothelial nitric oxide synthase) is responsible for the production of nitric oxide. eNOS is known to stimulate vasodilation and regulate inflammation and myocyte function. Impairment of endothelium-dependent vasodilatation that is observed in acute hyperglycaemia condition is due to suppression of ENOS. The gene might be involved in lipolysis. Variation in ENOS might lead to Henoch-Schönlein purpura. ENOS polymorphism affects the NO concentration in plasma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(-)-Epicatechin-induced recovery of mitochondria from simulated diabetes: Potential role of endothelial nitric oxide synthase.
Ramirez-Sanchez I
Diabetes & Vascular Disease Research : Official Journal of the International Society of Diabetes and Vascular Disease, 13(3), 201-210 (2016)
The role of the eNOS G894T and T-786C gene polymorphism in the development of ascites in cirrhosis.
Yildirim O
European Review for Medical and Pharmacological Sciences, 20(22), 4725-4730 (2016)
IFN-a Negatively Regulates the Expression of Endothelial Nitric Oxide Synthase and Nitric Oxide Production: Implications for Systemic Lupus Erythematosus.
Buie JJ
Journal of Immunology, 199(6), 1979-1988 (2017)
Functional G894T (rs1799983) polymorphism and intron-4 VNTR variant of nitric oxide synthase (NOS3) gene are susceptibility biomarkers of obesity among Tunisians.
Nasr HB
Obesity research & clinical practice, 10(4), 465-475 null
Mechanisms underlying the hypotensive and vasodilator effects of Ru(terpy)(bdq)NO](3+), a nitric oxide donor, differ between normotensive and spontaneously hypertensive rats.
Potje SR
European Journal of Pharmacology, 741, 222-229 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service