Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502011

Sigma-Aldrich

Anti-INOS antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HEP-NOS, hepatocyte NOS, inducible, inducible NOS, nitric oxide synthase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 131 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS2(4843)

Related Categories

General description

Anti-INOS antibody detects endogenous levels of total INOS protein.
The INOS (inducible nitric oxide synthase) gene is mapped to human chromosome 17q11.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human iNOS.

Immunogen Range: 117-166

Application

Anti-INOS, N-Terminal antibody produced in rabbit has been used in western blotting and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Upregulation of INOS (inducible nitric oxide synthase) is observed during infections, tissue injury, and inflammation. INOS is responsible to produce NO (nitric oxide), which controls blood flow by inducing vascular relaxation. NO acts as a neurotransmitter and can facilitate immune responses. NO can also bring about anti-thrombotic and anti-inflammatory effects in response to oxidative stress, hypoxia, inflammatory cytokines (TNF-α, IL-1β, IFN-γ), and lipopolysaccharide endotoxin. iNOS generates more NO than the neuronal and endothelial NOS. Unlike the two isoforms, iNOS is calcium independent. INOS might be associated with type 2 diabetes mellitus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Yang et al.
Molecular medicine reports, 13(2), 1220-1226 (2015-12-10)
Spinal nitric oxide is involved in the mechanisms of pain generation and transmission during inflammatory and neuropathic pain. The aim of the present study was to explore the role of spinal nitric oxide in the development of bone cancer pain.
Mateusz Adamiak et al.
Stem cell reviews, 13(1), 92-103 (2016-10-19)
Nitric oxide (NO) is a gaseous free radical molecule involved in several biological processes related to inflammation, tissue damage, and infections. Based on reports that NO inhibits migration of granulocytes and monocytes, we became interested in the role of inducible
R A Lea et al.
American journal of medical genetics, 105(1), 110-113 (2001-06-27)
Migraine is a debilitating disorder affecting approximately 12% of Caucasian populations. The disease has a large genetic component, although at present the type and number of genes involved is unclear. Candidate gene studies may be useful strategies for identifying genes
Feng Chen et al.
IUBMB life, 68(7), 516-525 (2016-05-20)
Inducible nitric oxide synthase (NOS2) and endothelial nitric oxide synthase (NOS3) gene play important roles in the susceptibility to type 2 diabetes mellitus (T2DM). The present study aims to detect the potential association of NOS2 and NOS3 gene polymorphisms with
Lu Chen et al.
International journal of oncology, 47(4), 1361-1370 (2015-09-01)
Vasoactive intestinal peptide (VIP) has been regarded as deactivator for macrophages. However, the depressive effect of VIP on tumor-associated macrophages (TAM) has not been recognized. In the present study, we investigated the effect of VIP on gastric cancer via TAM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service