Skip to Content
Merck
All Photos(5)

Key Documents

SAB4501894

Sigma-Aldrich

Anti-MMP-7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MMP-7, Matrilysin, Matrix metalloproteinase-7, Pump-1 protease, Uterine metalloproteinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 29 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP7(4316)

Related Categories

General description

Anti-MMP-7 Antibody detects endogenous levels of total MMP-7 protein.
Matrix metalloproteinase-7 (MMP7) is also termed as matrilysin. It is located on human chromosome 11q21-->q22. It belongs to the MMP enzyme family and is highly expressed in cancer cells. MMP7 is exclusively released by epithelial cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human MMP-7.

Immunogen Range: 218-267

Application

Anti-MMP-7, C-Terminal antibody has been used in immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Overexpression of matrix metalloproteinase-7 (MMP7) is observed in various cancers, such as breast, lung, prostate, esophagus, stomach, endometrium, and ovarian carcinomas, as well as esophageal squamous cell carcinomas. Activated MMP7 plays a vital role in human colorectal cancer (CRC) liver metastases. It is essential for the development of tumor in the absence of PKP3 (plakophilin3).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of MMP-7 Increases Collagen 1A2 in the Aging Kidney
Oelusarz A, et al.
Physiological Reports, 1(5) (2013)
Dan-Yang Yin et al.
Clinical kidney journal, 17(1), sfad027-sfad027 (2024-01-08)
To explore the advantages of urinary matrix metalloproteinase-7 (MMP-7) in evaluating renal tubular injury in minimal change disease (MCD) and focal segmental glomerulosclerosis (FSGS) patients compared with urinary cystatin C (CysC) and retinol-binding protein (RBP). Serum and urine samples were
Matrix metalloproteinase-7 expression in colorectal cancer liver metastases: evidence for involvement of MMP-7 activation in human cancer metastases.
Zeng ZS
Clinical Cancer Research, 8(1), 144-148 (2002)
Anna Oelusarz et al.
Physiological reports, 1(5) (2013-11-26)
The percentage of the U.S. population over 65 is rapidly increasing, as is the incidence of chronic kidney disease (CKD). The kidney is susceptible to age-dependent alterations in structure, specifically tubulointerstitial fibrosis, that lead to CKD. Matrix metalloproteinases (MMPs) were
Mapping of the metalloproteinase gene matrilysin (MMP7) to human chromosome 11q21-->q22.
Knox JD
Cytogenetics and Cell Genetics, 72(2-3), 179-182 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service