Skip to Content
Merck
All Photos(7)

Key Documents

HPA026761

Sigma-Aldrich

Anti-EPCAM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti Epcam, Anti-Epcam, Epcam Antibody, Epcam Antibody - Anti-EPCAM antibody produced in rabbit, Anti-CD326 antigen, Anti-EGP, Anti-Epithelial cell surface antigen, Anti-Epithelial glycoprotein, Anti-KS 1/4 antigen, Anti-KSA, Anti-Major gastrointestinal tumor-associated protein GA733-2, Anti-Tumor-associated calcium signal transducer 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LFKAKQCNGTSMCWCVNTAGVRRTDKDTEITCSERVRTYWIIIELKHKAREKPYDSKSLRTALQKEITTRYQLDPKFITSILYENNVITIDLVQNSSQKTQNDVDIADVAYY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPCAM(4072)

General description

Epithelial cell adhesion molecule (EpCAM) gene, also referred to as TACSTD (tumor-associated calcium signal transducer) 1, is mapped to human chromosome 2p21, a region composed of the mutS homolog 2 (MSH2) and mutS homolog 6 (MSH6) DNA mismatch repair genes. The gene codes for a membrane-associated glycoprotein expressed in epithelial cells of different organs.

Immunogen

Tumor-associated calcium signal transducer 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-EPCAM antibody produced in rabbit has been used in proximity ligation assay to study the interaction between EpCAM and AGR2 (anterior gradient-2 ) proteins.

Biochem/physiol Actions

Epithelial cell adhesion molecule (EpCAM) might function as a hepatic stem/progenitor cell-specific marker. EpCAM is an oncogene activated by deliverance of its intracellular domain, which is involved in transducing signals into the cell nucleus by associating with elements of the wnt pathway. Since the epithelial cell adhesion molecule (EpCAM) is highly expressed in premalignant hepatic tissues and in a subset of hepatocellular carcinoma (HCC), it might act as an early biomarker of hepatocellular carcinoma (HCC). EpCAM gene mutation leads to a rare autosomal recessive diarrheal disorder in infants called congenital tufting enteropathy (CTE). Like other cell adhesion molecules, EpCAM plays a crucial role in cell-cell interaction and it is also associated with tight junction protein, like claudin-7. Experimental studies hypothesize that EpCAM is involved in normal intestinal and crypt villus axis development .

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70142

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arshad Ayyaz et al.
Nature, 569(7754), 121-125 (2019-04-26)
The turnover of the intestinal epithelium is driven by multipotent LGR5+ crypt-base columnar cells (CBCs) located at the bottom of crypt zones1. However, CBCs are lost following injury, such as irradiation2, but the intestinal epithelium is nevertheless able to recover3.
The emerging role of EpCAM in cancer and stem cell signaling.
Cancer Research, 69(14), 5627-5629 (2009)
Absence of cell-surface EpCAM in congenital tufting enteropathy.
Schnell U
Human Molecular Genetics, 22(13), 2566-2571 (2013)
Assignment of TACSTD1 (alias TROP1, M4S1) to human chromosome 2p21 and refinement of mapping of TACSTD2 (alias TROP2, M1S1) to human chromosome 1p32 by in situ hybridization.
Calabrese G
Cytogenetics and Cell Genetics, 92(1-2), 164-165 (2001)
Min Su et al.
Stem cell research & therapy, 10(1), 239-239 (2019-08-08)
Type 1 diabetes (T1D) is an autoimmune disease resulting from the destruction of insulin-secreting islet β cells by autoreactive T cells. Non-obese diabetic (NOD) mice are the widely used animal model for human T1D. Autoimmunity in NOD mice is associated

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service