Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100139

Sigma-Aldrich

Anti-AR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AIS, Anti-Androgen receptor, Anti-DHTR, Anti-HUMARA, Anti-KD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

99 kDa

species reactivity

human, horse, dog, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AR(367)

Immunogen

Synthetic peptide directed towards the N terminal region of human AR

Biochem/physiol Actions

AR has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS).The androgen receptor gene is more than 90 kb long and codes for a protein that has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Two alternatively spliced variants encoding distinct isoforms have been described.

Sequence

Synthetic peptide located within the following region: MEVQLGLGRVYPRPPSKTYRGAFQNLFQSVREVIQNPGPRHPEAASAAPP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Tian et al.
Andrologia, 46(8), 859-866 (2013-10-16)
AR3, a major one of androgen receptor (AR) splice variants, has been shown to play a pivotal role in concert with AR signalling in prostate cancer. The present study was undertaken to characterise the expression pattern of AR3 in normal
James M Amos-Landgraf et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16514-16519 (2014-11-05)
It recently has been recognized that men develop colonic adenomas and carcinomas at an earlier age and at a higher rate than women. In the Apc(Pirc/+) (Pirc) rat model of early colonic cancer, this sex susceptibility was recapitulated, with male
Linda M Dairiki Shortliffe et al.
The Journal of urology, 191(6), 1913-1919 (2014-02-13)
Testosterone affects male development, maturation and aging but limited data exist on testosterone effects on the juvenile genitourinary system. We hypothesized that testosterone has bladder and kidney developmental effects, and investigated this in juvenile male rats. To examine the testosterone
Melissa A Babcook et al.
Molecular cancer therapeutics, 13(10), 2288-2302 (2014-08-15)
Docetaxel chemotherapy remains a standard of care for metastatic castration-resistant prostate cancer (CRPC). Docetaxel modestly increases survival, yet results in frequent occurrence of side effects and resistant disease. An alternate chemotherapy with greater efficacy and minimal side effects is needed.
Hung-Ming Lam et al.
Endocrine-related cancer, 21(6), 903-914 (2014-10-08)
Castration-resistant prostate cancer (CRPC) is an advanced-stage prostate cancer (PC) associated with high mortality. We reported that G-1, a selective agonist of G protein-coupled receptor 30 (GPR30), inhibited PC cell growth by inducing G2 cell cycle arrest and arrested PC-3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service