Skip to Content
Merck
All Photos(2)

Key Documents

A9853

Sigma-Aldrich

Anti-Androgen Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Androgen Receptor Antibody, Androgen Receptor Antibody - Anti-Androgen Receptor antibody produced in rabbit, Anti-AR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110 kDa (additional band may be observed at 45 kDa)

species reactivity

rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 20-40 μg/mL using microwave-pretreated human prostate tissue sections
immunoprecipitation (IP): 5-10 μg using human MCF7 cell lysate ( 250 μg)
microarray: suitable
western blot: 1-2 μg/mL using whole extract of rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AR(367)
rat ... Ar(24208)

General description

Androgens are the male sex hormones that exert biological and transcriptional effects on binding to nuclear androgen receptor (AR). Androgens in humans include androstenedione, testosterone, dihydrotestosterone and dehydroepiandrosterone. Androgen receptor is a ligand inducible nuclear receptor, expressed on brain, bone, skeletal muscles, genital organs, hair follicles and cardiovascular tissue. On binding androgen, the AR is internalized and modulates the transcriptional of target genes. Additionally, AR also exerts extra-nuclear actions wherein it rapidly induces signalling cascades of MAPK and protein kinase C. The regulation by AR results in irreversible sexual differentiation of sex organs during embryogenesis and development of sex-specific characteristics during puberty
Anti-Androgen Receptor specifically recognizes the human androgen receptor protein (110 kDa). An additional band may be observed at 45 kDa. The antibody cross-reacts with rat androgen receptor.

Specificity

The antibody cross-reacts with androgen receptor from rat.

Immunogen

evolutionally conserved peptide corresponding to amino acids 1-21 of the human androgen receptor. This sequence is not found in estrogen, progesterone, or glucocorticoid receptors.

Application

Anti-androgen receptor antibody may be used for immunoblotting using whole rat brain extract at a recommended concentration of 1-2 μg/ml. For immunoprecipitaion from 250 μg of human MCF7 cell lysate, a concentration of 5-10 μg may be used. Immunohistochemistry may be performed on paraffin-embedded human prostate tissue sections at a working antibody concentration of 20-40 μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Rabbit anti-androgen receptor (C19) was used in a EMSA (electrophoretic mobility shift assay) which produced a supershift in the molecular weight of a particular DNA sequence indicating binding of the androgen receptor. [1]

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura H Porter et al.
The Prostate, 79(11), 1326-1337 (2019-06-19)
Serially transplantable patient-derived xenografts (PDXs) are invaluable preclinical models for studying tumor biology and evaluating therapeutic agents. As these models are challenging to establish from prostate cancer specimens, the ability to preserve them through cryopreservation has several advantages for ongoing
D Orlic et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(18), 10344-10349 (2001-08-16)
Attempts to repair myocardial infarcts by transplanting cardiomyocytes or skeletal myoblasts have failed to reconstitute healthy myocardium and coronary vessels integrated structurally and functionally with the remaining viable portion of the ventricular wall. The recently discovered growth and transdifferentiation potential
Corrigan Horton et al.
Stem cell reports, 18(6), 1355-1370 (2023-05-13)
Both the canonical Wnt and androgen receptor (AR) signaling pathways are important for prostate organogenesis and homeostasis. How they crosstalk to regulate prostate stem cell behaviors remains unclear. Here, we show in lineage-tracing mouse models that although Wnt is essential
X Q Sun et al.
Journal of neuroendocrinology, 25(6), 547-559 (2013-02-19)
The seminal vesicles are male accessory sex glands that contribute much of the seminal fluid volume. Previous studies have suggested that the majority of autonomic innervations to the rat seminal vesicles originate from the bilateral major pelvic ganglia. Many preganglionic
Jou-Ching Lin et al.
Poultry science, 103(6), 103669-103669 (2024-04-12)
In the poultry industry, broiler and layer strains are genetically selected for different purposes (e.g., high meat-yield and high egg-production). Genetic selection for productivity can have unintended consequences on the behavioral repertoire of the birds, including aggression. Alongside the increasing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service