Merck
  • Synergistic simvastatin and metformin combination chemotherapy for osseous metastatic castration-resistant prostate cancer.
Synergistic simvastatin and metformin combination chemotherapy for osseous metastatic castration-resistant prostate cancer.
Molecular cancer therapeutics (2014-08-15)
Melissa A Babcook, Sanjeev Shukla, Pingfu Fu, Edwin J Vazquez, Michelle A Puchowicz, Joseph P Molter, Christine Z Oak, Gregory T MacLennan, Chris A Flask, Daniel J Lindner, Yvonne Parker, Firouz Daneshgari, Sanjay Gupta
ABSTRACT
Docetaxel chemotherapy remains a standard of care for metastatic castration-resistant prostate cancer (CRPC). Docetaxel modestly increases survival, yet results in frequent occurrence of side effects and resistant disease. An alternate chemotherapy with greater efficacy and minimal side effects is needed. Acquisition of metabolic aberrations promoting increased survival and metastasis in CRPC cells includes constitutive activation of Akt, loss of adenosine monophosphate-activated protein kinase (AMPK) activity due to Ser-485/491 phosphorylation, and overexpression of 3-hydroxy-3-methylglutaryl-Coenzyme A reductase (HMG-CoAR). We report that combination of simvastatin and metformin, within pharmacologic dose range (500 nmol/L to 4 μmol/L simvastatin and 250 μmol/L to 2 mmol/L metformin), significantly and synergistically reduces C4-2B3/B4 CRPC cell viability and metastatic properties, with minimal adverse effects on normal prostate epithelial cells. Combination of simvastatin and metformin decreased Akt Ser-473 and Thr-308 phosphorylation and AMPKα Ser-485/491 phosphorylation; increased Thr-172 phosphorylation and AMPKα activity, as assessed by increased Ser-79 and Ser-872 phosphorylation of acetyl-CoA carboxylase and HMG-CoAR, respectively; decreased HMG-CoAR activity; and reduced total cellular cholesterol and its synthesis in both cell lines. Studies of C4-2B4 orthotopic NCr-nu/nu mice further demonstrated that combination of simvastatin and metformin (3.5-7.0 μg/g body weight simvastatin and 175-350 μg/g body weight metformin) daily by oral gavage over a 9-week period significantly inhibited primary ventral prostate tumor formation, cachexia, bone metastasis, and biochemical failure more effectively than 24 μg/g body weight docetaxel intraperitoneally injected every 3 weeks, 7.0 μg/g/day simvastatin, or 350 μg/g/day metformin treatment alone, with significantly less toxicity and mortality than docetaxel, establishing combination of simvastatin and metformin as a promising chemotherapeutic alternative for metastatic CRPC.
Sigma-Aldrich
Anti-AR antibody produced in rabbit, affinity isolated antibody
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Supelco
Simvastatin, analytical standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard