Skip to Content
Merck
All Photos(1)

Documents

S2015

Sigma-Aldrich

SSPE Buffer 20× Concentrate

for Northern and Southern blotting, solution

Synonym(s):

20X SSPE Buffer, SSPE hybridization buffer, Saline-sodium phosphate-EDTA buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

solution

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
RNAse, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

SSPE Buffer 20x liquid is a standard reagent in Southern and Northern hybridization procedures. This liquid can be used straight or diluted as needed.

Application

Suitable for:
  • Hybridization buffer for Northern and Southern blots
  • Washing buffer for microarrays

Features and Benefits

  • Convenient ready-to-use liquid
  • Multi-purpose use in hybridizations

Components

SSPE Buffer 20x contains 0.02M EDTA and 2.98M NaCl in 0.2M phosphate buffer (pH 7.4).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide analysis of barcoded Saccharomyces cerevisiae gene-deletion mutants in pooled cultures.
Pierce SE, et al.
Nature Protocols, 2, 2958-2974 (2007)
Warren P Pickering et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 18(8), 1475-1484 (2003-08-05)
Increased protein degradation during metabolic acidosis contributes to muscle wasting in uraemia. Adrenalectomy experiments in severely acidotic rats (arterial pH approximately 7.15) have shown that this is prevented in the absence of glucocorticoid. It should therefore be possible to block
Glucocorticoid antagonist RU38486 fails to block acid-induced muscle wasting in vivo or in vitro.
Pickering WP, et al.
Nephrology, Dialysis, and Transplantation, 18, 1475-1484 (2003)
Claudia S Meier-Trummer et al.
PloS one, 4(7), e6265-e6265 (2009-07-16)
Malignant catarrhal fever (MCF) is a lethal disease of cattle, characterized by vasculitis, necrosis, and accumulation of activated, dysregulated cytotoxic lymphocytes in various tissues. Ovine gamma herpesvirus 2 (OvHV-2) is a causative agent of MCF, which may trigger the disease

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service