RNAINH
Murine RNase Inhibitor
suitable for cDNA synthesis, suitable for RT-PCR
About This Item
Recommended Products
form
liquid
Quality Level
concentration
40 unit/μL
technique(s)
PCR: suitable
RT-PCR: suitable
cDNA synthesis: suitable
qPCR: suitable
shipped in
wet ice
storage temp.
−20°C
General description
Compared with human-derived RNase inhibitors, mouse-derived RNase inhibitors do not contain two cysteines that are very sensitive to oxidation in human protein, so it has higher antioxidant activity and is more suitable for experiments sensitive to high DTT (such as qPCR).
This product is DNase, RNase and Nickase free.
Application
- in vitro inhibition of ribonucleases.
- cDNA synthesis.
- RT-PCR.
- in vitro transcription and translation.
Features and Benefits
RT-PCR/qPCR Compatibility:
- Enhanced antioxidant capacity will not form the disulfide bonds that human-derived proteins would produce under oxidative conditions.
- High-purity E. coli soluble expression.
- No RNase residue.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service