Skip to Content
Merck
All Photos(1)

Key Documents

R1280

Sigma-Aldrich

Monoclonal Anti-RPA/p34 antibody produced in mouse

~2 mg/mL, clone 9H8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-REPA2, Anti-RPA2, Anti-RPA32, Anti-Replication protein A2; 32kDa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human, monkey

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.1-0.2 μg/mL using HeLa total cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA2(6118)

General description

Monoclonal Anti-RPA/p34 (mouse IgG1 isotype) is derived from the hybridoma 9H8 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with purified human RPA/p34. Replication protein A Replication protein A (RPA) complex is a single-stranded DNA-binding protein and it is made up of 70-, 32- and 14-kDa subunits.

Immunogen

purified human RPA/p34.

Application

The antibody may be used in:
  • immunoblotting (~34 kDa)
  • immunocytochemistry
  • immunoprecipitation

Biochem/physiol Actions

Replication protein A (RPA) complex is essential for processes like DNA replication, recombination, and DNA repair pathways. The 32-kDa subunit of human RPA is hyper-phosphorylated in its N-terminus upon cellular DNA damage. This phosphorylation causes a change in RPA conformation that down-regulates its activity in DNA replication but does not affect DNA repair processes. Thus the role of RPA phosphorylation in the cellular response to DNA damage is to help regulate DNA metabolism and promote DNA repair.The RPA complex, once bound to single stranded DNA, recruits the ATR (ataxia-telangiectasia mutated and Rad3-related)-ATRIP (ATR-interacting protein) protein kinase complex that is crucial for the cellular response to replication stress and DNA damage and for ATR-mediated Chk1 activation in human cells. Furthermore, the association of this complex to the single stranded DNA causes the phosphorylation of RAD17 protein that is bound to DNA. Yeast strains deficient in RPA are defective in recruiting Ddc2 (yeast homologue of ATRIP) and thus down stream proteins are not phosphorylated. Replication protein A binds with the ssDNA with great affinity and interacts specifically with multiple proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Replication protein A phosphorylation and the cellular response to DNA damage
Binz S K, et al.
DNA Repair, 3(8-9), 1015-1024 (2004)
W Q Qiu et al.
The Journal of biological chemistry, 271(14), 8443-8451 (1996-04-05)
Progressive cerebral deposition of the amyloid beta-peptide (Abeta) is an early and constant feature of Alzheimer's disease. Abeta is derived by proteolysis from the beta-amyloid precursor protein. beta-Amyloid precursor protein processing and the generation of Abeta have been extensively characterized
Sensing DNA damage through ATRIP recognition of RPA-ssDNA complexes
Zou L and Elledge S J
Science (New York, N.Y.), 300(5625), 1542-1548 (2003)
M K Kenny et al.
The Journal of biological chemistry, 265(13), 7693-7700 (1990-05-05)
Human single-stranded DNA binding protein (human SSB) is a multisubunit protein containing polypeptides of 70, 34, and 11 kDa that is required for SV40 DNA replication in vitro. In this report we identify the functions of the SSB and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service