Skip to Content
Merck
All Photos(1)

Documents

D0720000

Dexamethasone sodium phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-phosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
Beilstein:
6473066
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dexamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O

InChI

1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1

InChI key

PLCQGRYPOISRTQ-FCJDYXGNSA-L

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone sodium phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veysel Nijat Baş et al.
European journal of pediatrics, 171(4), 735-736 (2011-12-29)
Iatrogenic Cushing syndrome may occur as an undesirable outcome of high-dose glucocorticoids treatments. This may also cause hypothalamus-hypophysis-adrenal axis suppression. While this situation may be caused more frequently with oral and topical glucocorticoid therapy, iatrogenic Cushing syndrome in childhood, caused
Martin Gräber et al.
ACS chemical biology, 6(10), 1008-1014 (2011-07-30)
Phosphorylation-dependent protein binding domains are crucially important for intracellular signaling pathways and thus highly relevant targets in chemical biology. By screening of chemical libraries against 12 structurally diverse phosphorylation-dependent protein binding domains, we have identified fosfosal and dexamethasone-21-phosphate as selective
Weina Liu et al.
Neuroscience letters, 524(2), 119-123 (2012-07-21)
Prenatal exposure to glucocorticoids (GCs) leads to affective dysfunction in adulthood, which may be associated with the alterations in hypothalamic-pituitary-adrenal (HPA) axis. Physical exercise has been shown to ameliorate depressive symptoms. The objectives of present study were to investigate whether
Anna Gonciarz et al.
Electrophoresis, 33(22), 3323-3330 (2012-10-26)
Plasma protein binding of drugs may have significant effect on its pharmacodynamic, toxicological and pharmacokinetic properties, since only the free drug can pass across biological membrane and get to its specific site of action. Many drugs show a high affinity
Amy E Cohen et al.
Ophthalmology, 119(1), 66-73 (2011-11-26)
Determine safe, effective, iontophoretic dose(s) of EGP-437 (dexamethasone phosphate formulated for iontophoresis) in patients with noninfectious anterior uveitis; evaluate systemic drug exposures. Prospective, phase I/II, multicenter, double-masked, parallel group, randomized clinical trial. Forty outpatients with anterior uveitis. Forty of 42

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service