Skip to Content
Merck

Skip To

IF014

Interferon-β Protein, Recombinant human

Recombinant Human Interferon Beta 1a (Hu-IFNbeta 1a).

Synonym(s):

Fibroblast Interferon

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Form:
liquid
Assay:
≥95% (one N-glycosylation site)
Biological source:
human
Recombinant:
expressed in CHO cells
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in CHO cells

assay

≥95% (one N-glycosylation site)

form

liquid

specific activity

3.99 x 108 U/mg

manufacturer/tradename

Chemicon®

concentration

1 x 106 units/mL

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

Product Source: cDNA obtained from human fibroblast mRNA expressed in CHO cells.
Recombinant Human Interferon Beta 1a (Hu-IFNbeta 1a). Molecular weight is ~23 kDa.

Physical form

Frozen liquid in 50 mM NaOAc, pH 5.5, containing 0.1% BSA.

Preparation Note

After receipt, this product should be kept at -70°C or below for retention of full activity.
Thaw product vial by incubation cold tap water until just thawed. Thawed IFNβ should be
stored in working aliquots at -70°C for up to six months. Avoid multiple freeze/thaw cycles.
Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the
product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).

Analysis Note

Specific Activity: Determined by assaying inhibition of virus-induced cytopathic effect.In this antiviral assay for interferon, about 1 unit/ml of interferon is the quantity necessary to produce a cytopathic effect of 50%.The units are determined with respect to the international reference standard for human interferon beta (Hu-IFNbeta) provided by the National Institutes of Health.Unit of activity measured on VERO cells with vesicular stomatitis virus (VSV).

Other Notes

Replaces: IF008

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
IF007IL038IF011
description

Recombinant Human Interferon Beta 1a (Hu-IFNbeta 1a).

description

Purified Recombinant Human Interferon Alpha A (Hu-IFN-alphaA & Hu-IFN-alpha2a).

description

The recombinant human Interleukin-1beta protein (or IL-1beta protein) is a potent immuno-modulator which mediates a wide range of immune & inflammatory responses including the activation of B & T-cells.

description

Interferon-β Protein, Recombinant mouse.

recombinant

expressed in CHO cells

recombinant

expressed in E. coli

recombinant

-

recombinant

expressed in E. coli

biological source

human

biological source

human

biological source

human

biological source

mouse

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

assay

≥95% (one N-glycosylation site)

assay

95%

assay

>98% (SDS-PAGE and HPLC)

assay

≥95%

concentration

1 x 106 units/mL

concentration

-

concentration

-

concentration

-

form

liquid

form

liquid

form

-

form

liquid


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters