Skip to Content
Merck
All Photos(1)

Key Documents

P3635

Sigma-Aldrich

5β-Pregnane-3α,20α-diol glucuronide

≥98.00% (TLC), powder, progesterone metabolite

Synonym(s):

P3G, PDG, Pn3-G, Pregnanediol-3a-glucuronide, Pregnanediol-glucuronide, 3α,20α-Dihydroxy-5β-pregnane 3-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O8
CAS Number:
Molecular Weight:
496.63
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Product Name

5β-Pregnane-3α,20α-diol glucuronide,

sterility

non-sterile

Assay

≥98.00% (TLC)

form

powder

technique(s)

enzyme immunoassay: suitable

solubility

ethanol: 20 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(O)C1CCC2C3CCC4CC(CCC4(C)C3CCC12C)OC5OC(C(O)C(O)C5O)C(O)=O

InChI

1S/C27H44O8/c1-13(28)17-6-7-18-16-5-4-14-12-15(8-10-26(14,2)19(16)9-11-27(17,18)3)34-25-22(31)20(29)21(30)23(35-25)24(32)33/h13-23,25,28-31H,4-12H2,1-3H3,(H,32,33)

InChI key

ZFFFJLDTCLJDHL-UHFFFAOYSA-N

General description

5β-Pregnane-3α,20α-diol glucuronide is a progesterone metabolite predominant in urine, during pregnancy. Presence of 5β-Pregnane-3α,20α-diol glucuronide in the urine indicates pregnancy, but it might also account for pseudopregnancy. It is useful in predicting the delivery time/end of pseudopregnancy.
Urinary pregnanediol glucuronide (PdG) seems to serve as an effective clinical marker for monitoring progesterone supplementation. PdG, along with other urinary metabolites such as luteinizing hormone (LH), follicle stimulating hormone (FSH), and estrone glucuronide (ElG), exhibit a strong association with ovarian excretion rates and demonstrate a high correlation with serum reproductive hormones. As a result, they can be used interchangeably to effectively monitor ovarian steroidogenesis.

Application

5β-Pregnane-3α,20α-diol glucuronide has been used in a cross-sectional analysis of hormone levels. It has also been used as a standard to measure estradiol-17β, pregnanediol-glucuronide (PdG) by a double-antibody enzyme immunoassay.
5β-Pregnane-3α,20α-diol glucuronide has been used in steroid enzyme immunoassay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chest Pain with Normal Coronary Arteries: A Multidisciplinary Approach, 323-323 (2013)
Jennifer E Mersereau et al.
Menopause (New York, N.Y.), 15(3), 482-486 (2008-01-19)
To compare daily reproductive hormone secretion in regularly menstruating older versus younger women. This was a prospective cohort study. Daily morning urine samples were obtained from 106 women, 28 of whom were aged between 20 and 34 years (mean: 27.8+/-3.7
Characterization of the ovarian cycle in the two-toed sloths (Choloepus didactylus): an innovative, reliable, and noninvasive method using fecal hormone analyses
Troll S, et al.
Theriogenology (2013)
Kathleen A O'Connor et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 18(3), 828-836 (2009-02-26)
Detailed characterization of estrogen dynamics during the transition to menopause is an important step toward understanding its potential implications for reproductive cancers developing in the transition years. We conducted a 5-year prospective study of endogenous levels of total and unopposed
Endocrine pregnancy monitoring in the two-toed sloth (Choloepus didactylus):?Pregnant or not pregnant?
Troll S, et al.
Theriogenology, 84(1), 137-144 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service