Skip to Content
Merck
All Photos(1)

Key Documents

N3415

Sigma-Aldrich

Nifurtimox

≥98% (HPLC)

Synonym(s):

(RS)-3-methyl-N-[(1E)-(5-nitro-2-furyl)methylene]thiomorpholin-4-amine 1,1-dioxide, (±)-Nifurtimox, 3-Methyl-4-(5′-nitrofurylidene-amino)-tetrahydro-4H-1,4-thiazine-1,1-dioxide; 4-((5-Nitrofurfurylidene)amino)-3-methylthiomorpholine 1,1-dioxide, BAY 2502, Thiomorpholine, 3-methyl-4-((5-nitrofurfurylidene)amino)-,1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O5S
CAS Number:
Molecular Weight:
287.29
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: ≥13 mg/mL

originator

Bayer

storage temp.

room temp

SMILES string

CC1CS(=O)(=O)CCN1\N=C\c2ccc(o2)[N+]([O-])=O

InChI

1S/C10H13N3O5S/c1-8-7-19(16,17)5-4-12(8)11-6-9-2-3-10(18-9)13(14)15/h2-3,6,8H,4-5,7H2,1H3/b11-6+

InChI key

ARFHIAQFJWUCFH-IZZDOVSWSA-N

General description

Nifurtimox acts as a hypoxia-activated cytotoxin, which specifically kills clonogenic tumor cells under hypoxic conditions. It is used to treat Chagas disease and African trypanosomiasis. Nifurtimox inhibits neuroblastoma and medulloblastoma cell growth.

Application

Nifurtimox has been used in drug treatment in culture media.

Biochem/physiol Actions

Nifurtimox is a nitrofurane derivative used to treat diseases caused by trypanosomes. Nifurtimox was discovered empirically and its mechanism of action is unclear. It is believed that nifurtimox exerts its biological activity through the bioreduction of the nitro-group to a nitro-anion radical which undergoes redox-cycling with molecular oxygen.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yvonne L Carter et al.
The American journal of tropical medicine and hygiene, 87(6), 1038-1040 (2012-10-24)
Acute Chagas disease is rarely recognized, and the risk for acquiring the disease is undefined in travelers to Central America. We describe a case of acute Chagas disease in a traveler to Costa Rica and highlight the need for increased
Hugo Cerecetto et al.
Future microbiology, 6(8), 847-850 (2011-08-25)
EVALUATION OF: Hall BS, Bot C, Wilkinson SR. Nifurtimox activation by trypanosomal type I nitroreductases generates cytotoxic nitrile metabolites. J. Biol. Chem. 286, 13088-13095 (2011). The prodrug nifurtimox has been one of the pharmacologic alternatives to treat Chagas disease and
P P Simarro et al.
Parasitology, 139(7), 842-846 (2012-02-09)
Despite the fact that eflornithine was considered as the safer drug to treat human African trypanosomiasis (HAT) and has been freely available since 2001, the difficulties in logistics and cost burden associated with this drug meant that the toxic melarsoprol
Sam Alsford et al.
Nature, 482(7384), 232-236 (2012-01-27)
The concept of disease-specific chemotherapy was developed a century ago. Dyes and arsenical compounds that displayed selectivity against trypanosomes were central to this work, and the drugs that emerged remain in use for treating human African trypanosomiasis (HAT). The importance
Monika Hułas-Stasiak et al.
Apoptosis : an international journal on programmed cell death, 16(10), 967-975 (2011-07-09)
This study was designed to determine follicular atresia in the newborn and the prepubertal spiny mouse. We analyzed the processes of follicle loss using classical markers of apoptosis (TUNEL reaction, active caspase-3) and autophagy (Lamp1). Numerous small clear vacuoles and

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service