Skip to Content
Merck
All Photos(1)

Key Documents

PHR1462

Supelco

Creatinine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Creatinine, 2-Amino-1-methyl-2-imidazolin-4-one, 2-Imino-1-methylimidazolidin-4-one, 2-Imino-N-methylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3O
CAS Number:
Molecular Weight:
113.12
Beilstein:
112061
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1150353

API family

creatinine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

295 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CN1CC(=O)N=C1N

InChI

1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)

InChI key

DDRJAANPRJIHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Creatinine is a by-product produced during the metabolism of creatine phosphate in muscle. It is excreted through urine, and serum creatinine level is most common indicator of renal health.

Application

Creatinine may be used as a pharmaceutical reference standard for the determination of the analyte in biological fluids by spectrophotometry, and high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3010 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

554.0 °F - closed cup

Flash Point(C)

290.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Creatinine
Clinical Veterinary Advisor, 615-615 (2013)
7.4 - Tubular Reabsorption and Secretion
Quantitative Human Physiology: An Introduction, 645-655 (2012)
Improved HPLC method for the simultaneous determination of allantoin, uric acid and creatinine in cattle urine
George SK
Journal of Chromatography. B, Biomedical Sciences and Applications, 832(1), 134-137 (2006)
A simplified method for HPLC determination of creatinine in mouse serum
Yuen PST, et al.
American Journal of Physiology: Renal Physiology, 286(6), F1116-F1119 (2004)
Creatinine determined by high-performance liquid chromatography.
Ambrose RT, et al.
Clinical Chemistry, 29(2), 256-259 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service