Skip to Content
Merck
All Photos(1)

Key Documents

P1512

Sigma-Aldrich

Thermolysin from Geobacillus stearothermophilus

Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)

Synonym(s):

Protease from Geobacillus stearothermophilus, Thermophilic-bacterial protease

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

Geobacillus stearothermophilus

Quality Level

type

Type X

form

lyophilized powder

specific activity

30-350 units/mg protein (E1%/280)

mol wt

34.6 kDa by amino acid sequence

purified by

crystallization

shipped in

wet ice

storage temp.

−20°C

General description

Thermolysin is a protease that has specificity different from other proteases available for sequence investigations.

Application

A thermostable (thermophilic) extracellular metalloendopeptidase containing four calcium ions. Cofactors are zinc and calcium. Hydrolyzes protein bonds on the N-terminal side of hydrophobic amino acid residues. The pH optimum is 8.0 and the optimal temperature for activity is 70 °C. Considerably stable from pH 5 to 9.5. Thermolysin has a low cleavage specificity, therefore, it produces a number of short fragments that are suitable for sequencing. Preferential cleavage: X-cleavage-Y-Z where X=any amino acid; Y=Leu, Phe, Ile, Val, Met, Ala and Z is any amino acid other than Pro. Cleavage N-terminal to Leu is preferred over cleavage of N-terminal to Phe which is preferred over the others. Often used to do limited proteolysis for peptide mapping and studies of protein structure and conformational changes.
Thermolysin has been shown to have a prosequeence that acts as an intramolecular chaperone in vivo. It has also been used in a study to investigate the effects of sodium chloride on thermal stability and catalytic activity.
Thermolysin is also commonly used for the commercial synthesis of N-(benzyloxycarbonyl)-L-aspartyl-L-phenylalanine methyl ester, the precursor for the artificial sweetener aspartame.

Quality

Contains many extraneous enzymes.

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Physical form

lyophilized powder containing calcium and sodium acetate buffer salts

Preparation Note

The pH optimum is 8.0 and the optimal temperature for activity is 70 °C. Considerably stable from pH 5 to 9.5. Thermolysin has a low cleavage specificity, therefore, it produces a number of short fragments that are suitable for sequencing. Preferential cleavage: X-cleavage-Y-Z where X=any amino acid; Y=Leu, Phe, Ile, Val, Met, Ala and Z is any amino acid other than Pro. Cleavage N-terminal to Leu is preferred over cleavage of N-terminal to Phe which is preferred over the others.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Miyanaga et al.
Biotechnology and bioengineering, 46(6), 631-635 (1995-06-20)
N-(benzyloxycarbonyl)-L-aspartyl-L-phenylalanine methyl ester, a precursor of the synthetic sweetener, aspartame, was synthesized from N-(benzyloxycarbonyl)-L-aspartic acid and L-phenylalanine methyl ester with an immobilized thermolysin (EC 3.4.24.4) in the mixed organic solvent system of tert-amyl alcohol and ethyl acetate. A mixed solvent
K Inouye et al.
Biochimica et biophysica acta, 1388(1), 209-214 (1998-10-17)
Thermolysin, a thermophilic metalloproteinase, is markedly activated in the presence of high concentrations (1-5 M) of neutral salts. The activity increases in an exponential fashion with increasing salt concentration, and is enhanced 13-15 times with 4 M NaCl at pH
The use of thermolysin in amino acid sequence determination.
R P Ambler et al.
The Biochemical journal, 108(5), 893-895 (1968-08-01)
Yongjin Park et al.
Scientific reports, 12(1), 10935-10935 (2022-06-30)
Long wavelengths that can deeply penetrate into human skin are required to maximize therapeutic effects. Hence, various studies on near-infrared organic light-emitting diodes (NIR OLEDs) have been conducted, and they have been applied in numerous fields. This paper presents a
Friedrich Hans Kleiner et al.
Journal of cell science, 134(9) (2021-09-23)
Cytochrome c6 is a redox carrier in the thylakoid lumen of cyanobacteria and some eukaryotic algae. Although the isofunctional plastocyanin is present in land plants and the green alga Chlamydomonas reinhardtii, these organisms also possess a cytochrome c6-like protein designated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service