Skip to Content
Merck
All Photos(1)

Key Documents

P3303

Sigma-Aldrich

Endoproteinase Asp-N from Pseudomonas fragi mutant strain

suitable for protein sequencing, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.56
Pricing and availability is not currently available.

grade

Proteomics Grade

Quality Level

form

lyophilized powder

analyte chemical class(es)

amino acids

packaging

vial of 2 μg

suitability

suitable for protein sequencing

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Endoproteinase Asp-N is a metallo endoprotease. It is obtained from a mutant strain of Pseudomonas fragi, which hydrolyzes peptide bonds on the N-terminal side of aspartic and cysteic acid residues.[1][2] Asp-N is used in proteomics for peptide mapping and protein sequence work due to its highly specific cleavage of peptides.[3][4]

Application

Endoproteinase Asp-N from Pseudomonas fragi mutant strain has been used for the digestion of specific proteins to prepare peptides and for the analysis of generated peptides by MS (mass spectrometry) method.[5][6][7]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional consequences of preorganized helical structure in the intrinsically disordered cell-cycle inhibitor p27(Kip1).
Bienkiewicz EA, et al.
Biochemistry, 41, 752-752 (2002)
Yeast ribosomal/cytochrome c SET domain methyltransferase subfamily: identification of Rpl23ab methylation sites and recognition motifs.
Kameoka D, et al.
Journal of Biochemistry, 134, 129-135 (2003)
S R Paik et al.
The Biochemical journal, 340 ( Pt 3), 821-828 (1999-06-09)
alpha-Synuclein is a component of the abnormal protein depositions in senile plaques and Lewy bodies of Alzheimer's disease (AD) and Parkinson's disease respectively. The protein was suggested to provide a possible nucleation centre for plaque formation in AD via selective
Sarah Murray et al.
Journal of virology, 80(12), 6171-6176 (2006-05-30)
Adeno-associated virus type 2 (AAV-2) capsid proteins have eight sequence motifs that are potential sites for O- or N-linked glycosylation. Three are in prominent surface locations, close to the sites of cellular receptor attachment and to neutralizing epitopes on or
Wilson KJ, et al.
Methods in Protein Sequence Analysis: Proceedings of the 7th International Conference, Berlin, July 3?8, 1988, 310-310 (1988)

Protocols

An optimized LC-MS/MS based workflow for low artifact tryptic digestion and peptide mapping of monoclonal antibody, adalimumab (Humira) using filter assisted sample preparation (FASP).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service