Skip to Content
Merck
All Photos(1)

Key Documents

P0021

Sigma-Aldrich

Pantoprazole sodium hydrate

≥98% (HPLC)

Synonym(s):

5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole sodium salt hydrate, Pantozol hydrate, Protonix hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F2N3O4S · Na · xH2O
CAS Number:
Molecular Weight:
405.35 (anhydrous basis)
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to off-white

solubility

H2O: ≥20 mg/mL

originator

Novartis

storage temp.

−20°C

SMILES string

O.[Na+].COc1ccnc(CS(=O)c2nc3cc(OC(F)F)ccc3[n-]2)c1OC

InChI

1S/C16H14F2N3O4S.Na.H2O/c1-23-13-5-6-19-12(14(13)24-2)8-26(22)16-20-10-4-3-9(25-15(17)18)7-11(10)21-16;;/h3-7,15H,8H2,1-2H3;;1H2/q-1;+1;

InChI key

CGJRLPRCWSHOFU-UHFFFAOYSA-N

Gene Information

Application

Pantoprazole sodium hydrate has been used:
  • as a standard in high-performance liquid chromatography (HPLC){58
  • as a prazole to study its effects on prodrug activation and human immunodeficiency virus 1 (HIV-1) inhibition
  • to study its effects on breast cancer resistance protein (BCRP) inhibitors in various in vitro membrane preparations from various cell lines

Biochem/physiol Actions

Pantoprazole has the potential to treat symptoms of heartburn and acid regurgitation. It also has been studied to treat Helicobacter pylori infection and duodenal ulcer.
Pantoprazole is a benzimidazole derivative,activated in acidic environment. It is useful in treating long term prophylaxis and acid-related disorders.
Pantoprazole is a gastric proton pump inhibitor.

Features and Benefits

This compound is featured on the Acid-Sensing (Proton-gated) Ion Channels (ASICs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fitton et al.
Drugs, 51(3), 460-482 (1996-03-01)
Pantoprazole is an irreversible proton pump inhibitor which, at the therapeutic dose of 40mg, effectively reduces gastric acid secretion. In controlled clinical trials, pantoprazole (40mg once daily) has proved superior to ranitidine (300mg once daily or 150mg twice daily) and
Analysis of ibuprofen, pantoprazole, and itopride combination therapeutic drugs in human plasma by solid phase membrane microtip extraction and high-performance liquid chromatography methods using new generation core shell C18 column
Ali I, et al.
Journal of Liquid Chromatography and Related Technologies, 39(7), 339-345 (2016)
<BIG>Rissling O</BIG>
Interaction of Mycophenolic Acid and Pantoprazole: A Pharmacokinetic Crossover Study (2017)
Pantoprazole
<BIG>Perry CM and Ibbotson T</BIG>
Drugs, 101-132 (2003)
Maryam H Shubbar et al.
European journal of pharmacology, 874, 173009-173009 (2020-02-18)
Deposition of amyloid-β peptide (Aβ(1-42)) is a hallmark of Alzheimer's disease. Clearance of Aβ(1-42), across the blood-brain barrier (BBB), is mediated by ATP-binding Cassette (ABC) efflux transporters. Many therapeutic drugs inhibit ABC transporters, but little is known of the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service