Skip to Content
Merck
All Photos(1)

Key Documents

D2950000

Disulfiram

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tetraethylthiuram disulfide, Bis(diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamyl) disulfide, Disulfiram, TETD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSS2CSN(C2H5)2
CAS Number:
Molecular Weight:
296.54
Beilstein:
1712560
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

disulfiram

manufacturer/tradename

EDQM

mp

69-71 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCN(CC)C(=S)SSC(=S)N(CC)CC

InChI

1S/C10H20N2S4/c1-5-11(6-2)9(13)15-16-10(14)12(7-3)8-4/h5-8H2,1-4H3

InChI key

AUZONCFQVSMFAP-UHFFFAOYSA-N

Gene Information

human ... ALDH2(217)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alcohol dehydrogenase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Cvek et al.
Drug discovery today, 13(15-16), 716-722 (2008-06-27)
The major approach to the development of anticancer drugs involves searching for new compounds, efficient against malignancies, which are not, as yet, used clinically. This strategy is time-consuming and expensive. Recent studies have disclosed a surprising, but mechanistically consistent, anticancer
Meriem Gaval-Cruz et al.
Molecular interventions, 9(4), 175-187 (2009-09-02)
The anti-alcoholism drug disulfiram (Antabuse), which is an inhibitor of aldehyde dehydrogenase, induces an aversive reaction to alcohol consumption and thereby helps patients reduce alcohol intake. Recent clinical trials, initiated to investigate whether disulfiram could be used to treat individuals
Colin Rae et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 953-960 (2013-04-26)
Disulfiram has been used for several decades in the treatment of alcoholism. It now shows promise as an anticancer drug and radiosensitizer. Proposed mechanisms of action include the induction of oxidative stress and inhibition of proteasome activity. Our purpose was
Parvinder Hothi et al.
Oncotarget, 3(10), 1124-1136 (2012-11-21)
Glioblastoma Multiforme (GBM) continues to have a poor patient prognosis despite optimal standard of care. Glioma stem cells (GSCs) have been implicated as the presumed cause of tumor recurrence and resistance to therapy. With this in mind, we screened a
Thomas R Kosten et al.
Biological psychiatry, 73(3), 219-224 (2012-08-22)
Disulfiram has been an effective cocaine addiction pharmacotherapy, and one of its possible mechanisms of efficacy is through copper chelation and inhibition of an enzyme involved in catecholamine metabolism, dopamine β-hydroxylase (DβH), which converts dopamine to norepinephrine. A variant in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service