Skip to Content
Merck
All Photos(1)

Documents

L1751

Sigma-Aldrich

Ethyl linoleate

≥99%

Synonym(s):

Linoleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)2(CH2)7COOC2H5
CAS Number:
Molecular Weight:
308.50
Beilstein:
1727827
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant oil (safflower)

Assay

≥99%

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

224 °C/17 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h8-9,11-12H,3-7,10,13-19H2,1-2H3/b9-8-,12-11-

InChI key

FMMOOAYVCKXGMF-MURFETPASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of self-microemulsifying drug delivery system for oral bioavailability enhancement of berberine hydrochloride.: This study explores the formulation of a self-microemulsifying drug delivery system (SMEDDS) using ethyl linoleate as a key component to improve the oral bioavailability of berberine hydrochloride. The findings indicate a significant increase in bioavailability, demonstrating the potential of SMEDDS in enhancing the absorption of poorly water-soluble drugs (Zhu et al., 2013).

  • Metabonomic investigation of liver profiles of nonpolar metabolites obtained from alcohol-dosed rats and mice using high mass accuracy MSn analysis.: The research includes the use of ethyl linoleate in the study of liver metabolomics to understand the biochemical effects of alcohol dosing. Ethyl linoleate′s role in nonpolar metabolite profiling provides insights into liver function and the metabolic impacts of alcohol consumption (Loftus et al., 2011).

  • Enhanced bioavailability of silymarin by self-microemulsifying drug delivery system.: This article reports on the utilization of ethyl linoleate in a self-microemulsifying drug delivery system to enhance the bioavailability of silymarin. The study shows improved absorption and bioavailability of silymarin, highlighting the effectiveness of ethyl linoleate in drug delivery applications (Wu et al., 2006).

Biochem/physiol Actions

Ethyl linoleate may be used as a reference material in assays that quantify fatty acid ethyl esters (FAEEs) for detection of alcohol abuse. Linoleic acid ethyl ester (Ethyl linoleate) may be used and studied as an acetylcholinesterase (AChE) inhibitor.
Ethyl linoleate solated from Oxalis triangularis inhibits forskolin-induced melanogenesis and tyrosinase activity in mouse B16 melanoma cells . This anti-melanogenic effect is mediated by inhibiting cAMP production.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janine R Hutson et al.
Alcohol (Fayetteville, N.Y.), 45(2), 193-199 (2010-08-14)
Fatty acid ethyl esters (FAEEs) are nonoxidative metabolites of ethanol, and elevated levels of FAEE in meconium are a useful biomarker for heavy prenatal alcohol exposure. FAEE in meconium has been recommended as useful and cost-effective for universal screening for
Jonathan Kissling et al.
Phytotherapy research : PTR, 19(11), 984-987 (2005-12-01)
In a search for potential acetylcholinesterase (AChE) inhibitors, the ethanol extract of the bulbs of Crinum x powellii (Amaryllidaceae) was found to demonstrate a marked inhibition of this enzyme. Using a bio-guided isolation strategy, linoleic acid ethyl ester has been
Abdulsallam Bakdash et al.
Analytical and bioanalytical chemistry, 396(7), 2469-2477 (2010-02-11)
Fatty acid ethyl esters (FAEE) and ethyl glucuronide (EtG) were determined in 602 meconium samples in a maternal health evaluation study for detection of gestational alcohol consumption. A validated headspace solid phase microextraction method in combination with GC-MS was used
Fritz Pragst et al.
Therapeutic drug monitoring, 30(2), 255-263 (2008-03-28)
The retrospective detection of alcohol consumption during pregnancy is an important part of the diagnosis of the fetal alcohol syndrome. A promising way to solve this problem can be the determination of fatty acid ethyl esters (FAEE) or/and ethyl glucuronide
Ho-Seok Kwak et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(21), 1871-1874 (2010-07-08)
This paper reports the development and validation of an improved assay for quantitation of fatty acid ethyl esters (FAEEs) in human meconium using liquid chromatography/tandem mass spectrometry (LC-MS/MS). FAAEs (ethyl laurate, ethyl myristate, ethyl palmitate, ethyl palmitoleate, ethyl stearate, ethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service