Skip to Content
Merck
All Photos(2)

Documents

O6877

Sigma-Aldrich

Octanoyl coenzyme A lithium salt hydrate

≥95% (HPLC)

Synonym(s):

Capryloyl coenzyme A lithium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50N7O17P3S · xLi+ · yH2O
CAS Number:
Molecular Weight:
893.73 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].O.CCCCCCCC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)([O-])=O)n2cnc3c(N)ncnc23

InChI

1S/C29H50N7O17P3S.3Li.H2O/c1-4-5-6-7-8-9-20(38)57-13-12-31-19(37)10-11-32-27(41)24(40)29(2,3)15-50-56(47,48)53-55(45,46)49-14-18-23(52-54(42,43)44)22(39)28(51-18)36-17-35-21-25(30)33-16-34-26(21)36;;;;/h16-18,22-24,28,39-40H,4-15H2,1-3H3,(H,31,37)(H,32,41)(H,45,46)(H,47,48)(H2,30,33,34)(H2,42,43,44);;;;1H2/q;3*+1;/p-3/t18-,22-,23-,24+,28-;;;;/m1..../s1

InChI key

VWXPWBSHKDXAFH-SMMZWZJBSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Octanoyl coenzyme A is a donor molecule for the acylation (octanylation) of Ghrelin and other peptides by ghrelin O-acyltransferase(s) (GOAT).

Biochem/physiol Actions

Medium-chain fatty acid covalently linked to coenzyme A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The expression of ghrelin O-acyltransferase (GOAT) in human tissues.
Lim CT, Kola B, Grossman A, Korbonits M.
Endocrine J., 58, 707-710 (2011)
Amparo Romero et al.
European journal of endocrinology, 163(1), 1-8 (2010-04-28)
The ghrelin-ghrelin receptor system is one of the most important mechanisms regulating energy balance and metabolism. Among other actions, central and peripheral administration of ghrelin increases food intake and adiposity. During the last years, many efforts have been made in
Jesus A Gutierrez et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(17), 6320-6325 (2008-04-30)
The peptide hormone ghrelin is the only known protein modified with an O-linked octanoyl side group, which occurs on its third serine residue. This modification is crucial for ghrelin's physiological effects including regulation of feeding, adiposity, and insulin secretion. Despite
Tomoko Takahashi et al.
Journal of biochemistry, 146(5), 675-682 (2009-07-25)
Ghrelin was originally isolated from rat stomach as an endogenous ligand for the GH secretagogue receptor. The major active form of ghrelin is a 28-amino acid peptide modified by an n-octanoic acid on the serine 3 residue, and this lipid
Jing Yang et al.
Cell, 132(3), 387-396 (2008-02-13)
Ghrelin is a 28 amino acid, appetite-stimulating peptide hormone secreted by the food-deprived stomach. Serine-3 of ghrelin is acylated with an eight-carbon fatty acid, octanoate, which is required for its endocrine actions. Here, we identify GOAT (Ghrelin O-Acyltransferase), a polytopic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service