Skip to Content
Merck
All Photos(1)

Key Documents

870716P

Avanti

16:0 Coenzyme A

Avanti Research - A Croda Brand 870716P, powder

Synonym(s):

palmitoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H75N10O17P3S
CAS Number:
Molecular Weight:
1057.03
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (870716P-10mg)
pkg of 1 × 25 mg (870716P-25mg)
pkg of 1 × 5 mg (870716P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870716P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C37H66N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-28(46)65-21-20-39-27(45)18-19-40-35(49)32(48)37(2,3)23-58-64(55,56)61-63(53,54)57-22-26-31(60-62(50,51)52)30(47)36(59-26)44-25-43-29-33(38)41-24-42-34(29)44;;;/h24-26,30-32,36,47-48H,4-23H2,1-3H3,(H,39,45)(H,40,49)(H,53,54)(H,55,56)(H2,38,41,42)(H2,50,51,52);3*1H3/t26-,30?,31+,32+,36-;;;/m1.../s1

InChI key

VJXJWGKNGIGUQX-AUCFWDBISA-N

General description

16:0 Coenzyme A, also called as palmitoyl coenzyme A, is a most abundant acyl-CoA and the product of the first round of β-oxidation. It serves as a substrate for palmitoylation, a process in which a protein acyltransferase transfers the palmitoyl group to a free thiol on target proteins.

Application

16:0 Coenzyme A has been used as a:
  • standard in mass spectrometry for qualitative analysis of palmitoyl acyl-CoA in mycobacterium cells
  • constituent of the binding buffer in protein in vitro binding assay
  • substrate in serine palmitoyltransferase assay

Packaging

5 mL Amber Glass Screw Cap Vial (870716P-10mg)
5 mL Amber Glass Screw Cap Vial (870716P-25mg)
5 mL Amber Glass Screw Cap Vial (870716P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheryl Budde et al.
Methods (San Diego, Calif.), 40(2), 143-150 (2006-10-03)
Palmitoylation enhances membrane association and plays a role in the subcellular trafficking and signaling function of proteins. Unlike other forms of protein lipidation, such as prenylation and myristoylation, palmitoylation is reversible and can therefore play a regulatory role. Enzyme activities
Analysis of coenzyme A activated compounds in actinomycetes
Cabruja M, et al.
Applied Microbiology and Biotechnology, 100(16), 7239-7248 (2016)
Matías Cabruja et al.
Applied microbiology and biotechnology, 100(16), 7239-7248 (2016-06-09)
Acyl-CoAs are crucial compounds involved in essential metabolic pathways such as the Krebs cycle and lipid, carbohydrate, and amino acid metabolisms, and they are also key signal molecules involved in the transcriptional regulation of lipid biosynthesis in many organisms. In
Oshrit Ben-David et al.
The Journal of biological chemistry, 286(34), 30022-30033 (2011-06-28)
Sphingolipids (SLs) act as signaling molecules and as structural components in both neuronal cells and myelin. We now characterize the biochemical, histological, and behavioral abnormalities in the brain of a mouse lacking very long acyl (C22-C24) chain SLs. This mouse
Jeong-Won Nam et al.
The Plant cell, 32(4), 820-832 (2020-02-16)
Acyl carrier proteins (ACPs) are the scaffolds for fatty acid biosynthesis in living systems, rendering them essential to a comprehensive understanding of lipid metabolism. However, accurate quantitative methods to assess individual acyl-ACPs do not exist. We developed a robust method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service