O1012
Oleoyl coenzyme A lithium salt
≥90% (HPLC)
Synonym(s):
cis-9-Octadecenoyl coenzyme A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C39H68N7O17P3S
CAS Number:
Molecular Weight:
1031.98
UNSPSC Code:
41106305
NACRES:
NA.51
Recommended Products
Assay
≥90% (HPLC)
form
powder
solubility
H2O: soluble-50 mg/mL, clear, colorless
storage temp.
−20°C
Application
Coenzyme A functions as an acyl group carrier, acetyl-CoA. Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.
Biochem/physiol Actions
Oleoyl-CoA is an important substrate, that is essential for the biosynthesis of 1-Alkyl-2,3-diacylglycerol (ADG) and very long chain monounsaturated fatty acids of harderian gland (HG) 1-alkyl-2,3-diacylglycerol.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ta-Yuan Chang et al.
American journal of physiology. Endocrinology and metabolism, 297(1), E1-E9 (2009-01-15)
The enzymes acyl-coenzyme A (CoA):cholesterol acyltransferases (ACATs) are membrane-bound proteins that utilize long-chain fatty acyl-CoA and cholesterol as substrates to form cholesteryl esters. In mammals, two isoenzymes, ACAT1 and ACAT2, encoded by two different genes, exist. ACATs play important roles
Chengcheng Guan et al.
Nature communications, 11(1), 2478-2478 (2020-05-20)
Sterol O-acyltransferase 1 (SOAT1) is an endoplasmic reticulum (ER) resident, multi-transmembrane enzyme that belongs to the membrane-bound O-acyltransferase (MBOAT) family. It catalyzes the esterification of cholesterol to generate cholesteryl esters for cholesterol storage. SOAT1 is a target to treat several
Potential role of acyl-coenzyme A:cholesterol transferase (ACAT) Inhibitors as hypolipidemic and antiatherosclerosis drugs.
Leon C, Hill JS, Wasan KM.
Pharmaceut. Res., 22, 1578-1588 (2005)
Oleoyl-CoA is the major de novo product of stearoyl-CoA desaturase 1 gene isoform and substrate for the biosynthesis of the Harderian gland 1-alkyl-2, 3-diacylglycerol
Miyazaki M, et al.
The Journal of Biological Chemistry, 276(42), 39455-39461 (2001)
Mariana Igoillo-Esteve et al.
International journal for parasitology, 41(3-4), 429-438 (2010-12-18)
Metabolism in trypanosomatids is compartmentalised with major pathways, notably glycolysis, present in peroxisome-like organelles called glycosomes. To date, little information is available about the transport of metabolites through the glycosomal membrane. Previously, three ATP-binding cassette (ABC) transporters, called GAT1-3 for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service