Skip to Content
Merck
All Photos(1)

Documents

N6514

Sigma-Aldrich

Neuraminidase from Vibrio cholerae

Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)

Synonym(s):

Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Vibrio cholerae

Quality Level

type

Type II

form

buffered aqueous solution

specific activity

8-24 units/mg protein (Lowry, using NAN-lactose)

foreign activity

Protease and NAN-aldolase, present

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Calorimetry studies show that the two lectin-like domains flanking the central catalytic domain of Neuraminidase serve as the recognition and binding sites for sialic acid-containing substrates.
Neuraminidase enzymes are hydrolase enzymes that promote influenza virus release from infected cells and facilitate virus spread.

Application

Neuraminidase from Vibrio cholera has been used in a study to describe a five-step purification method. It has also been used in a study to investigate modification of leukemia L1210 tumor cells.
Neurminidase is used as a cell-surface probe for glycoconjugate distribution and in substrate specificity studies.

Quality

Preservative free.

Unit Definition

One unit will liberate 1.0 μmole of N-acetylneuraminic acid per min at pH 5.0 at 37 °C using NAN-lactose or bovine submaxillary mucin, unless otherwise specified. Prices based on units using NAN-lactose as substrate.

Physical form

Solution in 50 mM sodium acetate, pH 5.5, containing 0.15 M sodium chloride and 4 mM calcium chloride, 0.2 μm filtered..

Preparation Note

A further purification by affinity chromatography of our Type III.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Moustafa et al.
The Journal of biological chemistry, 279(39), 40819-40826 (2004-07-01)
Vibrio cholerae neuraminidase (VCNA) plays a significant role in the pathogenesis of cholera by removing sialic acid from higher order gangliosides to unmask GM1, the receptor for cholera toxin. We previously showed that the structure of VCNA is composed of
S Crennell et al.
Structure (London, England : 1993), 2(6), 535-544 (1994-06-15)
Vibrio cholerae neuraminidase is part of a mucinase complex which may function in pathogenesis by degrading the mucin layer of the gastrointestinal tract. The neuraminidase, which has been the target of extensive inhibitor studies, plays a subtle role in the
Increase of leukemia L1210 immunogenicity by Vibrio cholerae neuraminidase treatment.
J G Bekesi et al.
Cancer research, 31(12), 2130-2132 (1971-12-01)
Paul Clark et al.
Viruses, 12(10) (2020-10-24)
Polyomaviruses are small, non-enveloped DNA tumor viruses that cause serious disease in immunosuppressed people, including progressive multifocal leukoencephalopathy (PML) in patients infected with JC polyomavirus, but the molecular events mediating polyomavirus entry are poorly understood. Through genetic knockdown approaches, we
Hirokazu Kai et al.
Bioorganic & medicinal chemistry, 20(8), 2739-2746 (2012-03-14)
A selective and potent inhibitor of neuraminidases, a hydrolase that is responsible for processing sialylated glycoconjugates, is a promising drug candidate for various infective diseases. The current study demonstrates that the use of an aglycone-focused library of 2-difluoromethylphenyl α-sialosides is

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Protocols

Enzymatic Assay of Neuraminidase applies to products that have a specification for neuraminidase content by enzymatic determination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service