Skip to Content
Merck

A4789

Anti-Mouse IgA (α-chain specific)−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgA (α-chain specific)−HRP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
peroxidase conjugate
Clone:
polyclonal
Application:
ELISA (d)
Citations:
28
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgA antibody plays an essential role in mucosal immunity and binds with immunogens/pathogens to restrict them from entering the mucosal membrane. Anti-mouse IgA (α-chain specific)-peroxidase antibody can be used for determining the secretory IgA (sIgA) in intestinal fluid. It can also be used in direct ELISA. Goat anti-mouse IgA (α-chain specific)-peroxidase antibody reacts specifically with mouse IgA.

Immunogen

Purified mouse IgA

Application

Anti-mouse IgA (α-chain specific)-peroxidase antibody can be used in modified indirect DIBA (dot- immunobinding assay) .
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Western Blotting (1 paper)
ELISAS were performed using HRP conjugated gaot anti-mouse IgA as the secondary antibody at a dilution of 1:10000 for a 1 hour incubation at 37 degrees to detect IgA levels in mouse fecal extracts.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p 215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library