Skip to Content
Merck
All Photos(1)

Key Documents

N7271

Sigma-Aldrich

α(2→3) Neuraminidase from Streptococcus pneumoniae

buffered aqueous solution

Synonym(s):

Acyl-neuraminyl Hydrolase, Acylneuraminyl hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

Streptococcus pneumoniae

Quality Level

form

buffered aqueous solution

concentration

≥5 units/mL

foreign activity

β-Galactosidase, α-mannosidase, β-hexosaminidase, α-fucosidase, and proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Streptococcus pneumoniae R6 ... nanB(933504)

Biochem/physiol Actions

Releases α(2→3)-linked N-acetylneuraminic acid from complex oligosaccharides.

Packaging

Provided with 5× reaction buffer (250 mM sodium phosphate, pH 6.0).

Unit Definition

One unit will hydrolyze 1μmole of 4-methylumbelliferyl α-D-N-acetylneuraminide per min at pH 5.0 at 37 °C.

Physical form

Solution in 50 mM sodium phosphate, pH 7.5

Preparation Note

Expressed in glycosidase-free hosts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominic Meusch et al.
Nature, 508(7494), 61-65 (2014-02-28)
Tripartite Tc toxin complexes of bacterial pathogens perforate the host membrane and translocate toxic enzymes into the host cell, including in humans. The underlying mechanism is complex but poorly understood. Here we report the first, to our knowledge, high-resolution structures
Johan Nordholm et al.
The Journal of biological chemistry, 288(15), 10652-10660 (2013-03-01)
Interactions that facilitate transmembrane domain (TMD) dimerization have been identified mainly using synthetic TMDs. Here, we investigated how inherent properties within natural TMDs modulate their interaction strength by exploiting the sequence variation in the nine neuraminidase subtypes (N1-N9) and the
Audu J Natala et al.
Journal of medical entomology, 50(1), 85-93 (2013-02-23)
Amblyomma variegatum F. are obligate hematophagous ectoparasites of livestock that serve as the vectors of Ehrlichia ruminantium (formerly known as Cowdria ruminantium), the causative agent of heartwater disease. In the light of the fact that they are blood-feeding, their salivary
Nenad Macesic et al.
The Medical journal of Australia, 198(7), 370-372 (2013-04-16)
To review cases of nosocomial influenza and compare the epidemiology, clinical characteristics and outcomes with community-acquired cases. Prospective case series of adults hospitalised with influenza during April - November of 2010 and 2011 using a hospital-based sentinel surveillance system. A
Michael Worobey et al.
Nature, 508(7495), 254-257 (2014-02-18)
Zoonotic infectious diseases such as influenza continue to pose a grave threat to human health. However, the factors that mediate the emergence of RNA viruses such as influenza A virus (IAV) are still incompletely understood. Phylogenetic inference is crucial to reconstructing

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service