Skip to Content
Merck
All Photos(1)

Key Documents

N6394

Sigma-Aldrich

cis-10-Nonadecenoic acid

≥99%, liquid

Synonym(s):

(Z)-10-Nonadecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

liquid

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCC(O)=O

InChI

1S/C19H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21/h9-10H,2-8,11-18H2,1H3,(H,20,21)/b10-9-

InChI key

BBOWBNGUEWHNQZ-KTKRTIGZSA-N

Packaging

Sealed ampule.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dandan Feng et al.
Oncotarget, 8(55), 94692-94710 (2017-12-08)
Xuefu Zhuyu Decoction (XFZY), an important traditional Chinese herbal formula, has been reported effective on traumatic brain injury (TBI) in rats. However, its cerebral protection mechanism has not been clarified at the metabolic level. This work aims to explore the
Hui-Hui Xiao et al.
Frontiers in pharmacology, 9, 932-932 (2018-09-07)
The lignan-rich fraction (SWR) of Sambucus Williamsii Ramulus, a folk herbal medicine in China for treatment of bone diseases, has previously reported to exert protective effects on bone without exerting uterotrophic effects in ovariectomized (OVX) mice. The aim of the
Ling Dai et al.
Talanta, 135, 108-114 (2015-02-03)
Metabolic syndrome (MetS) is a cluster of metabolic abnormalities associated with an increased risk of developing cardiovascular diseases or type II diabetes. Till now, the etiology of MetS is complex and still unknown. Metabolic profiling is a powerful tool for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service