SRE0027
Heparin sodium porcine mucosa
Synonym(s):
Heparin sodium salt from porcine intestinal mucosa
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Porcine (mucosa)
Quality Level
form
powder
application(s)
diagnostic assay manufacturing
Looking for similar products? Visit Product Comparison Guide
Other Notes
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
recommended
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of cellular biochemistry, 118(9), 3003-3015 (2017-03-03)
In vitro assembly of functional liver tissue is a prerequisite for the transplantation of tissue-engineered livers. There is an increasing demand for in vitro models that replicate complex events occurring in the liver. However, tissue engineering of implantable liver systems
Nature, 513(7518), 436-439 (2014-07-22)
Fibroblast growth factor 1 (FGF1) is an autocrine/paracrine regulator whose binding to heparan sulphate proteoglycans effectively precludes its circulation. Although FGF1 is known as a mitogenic factor, FGF1 knockout mice develop insulin resistance when stressed by a high-fat diet, suggesting a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service