Skip to Content
Merck
All Photos(1)

Key Documents

58983

Supelco

SUPELCOSIL LC-8 (3 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-8 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L7

Quality Level

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

6.0% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.2 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C8 (octyl) phase

particle size

3 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

A phase less hydrophobic than C18. Provides less retention of both polar and non-polar compounds than C18. Use a mobile phase containing 5% less organic modifier for the C8 column than C18. Polar compounds are, relatively, more strongly retained on C8 than C18 columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Borch et al.
Journal of chromatography. A, 1022(1-2), 83-94 (2004-02-03)
Explosives such as 2,4,6-trinitrotoluene (TNT), octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX), and hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) are widely distributed environmental contaminants. Complete chromatographic separation is necessary in order to accurately determine and quantify explosives and their degradation products in environmental samples and in (bio)transformation studies. The
F Belal et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 335-342 (2001-02-24)
A simple, rapid and sensitive HPLC method has been developed for the simultaneous determination of ramipril and hydrochlorothiazide in their dosage forms. Acetonitrile: sodium perchlorate solution (0.1 M) adjusted to pH 2.5+/-0.2 with phosphoric acid (46:54 v/v), was used as
Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
K Ary et al.
Journal of pharmaceutical and biomedical analysis, 26(2), 179-187 (2001-07-27)
A reversed-phase high-performance liquid chromatographic method with coulometric and UV detection has been developed for the simultaneous determination of morphine, morphine-3-glucuronide and morphine-6-glucuronide. The separation was carried out by using a Supelcosil LC-8 DB reversed-phase column and 0.1 M potassium
Vesna Cerkvenik-Flajs et al.
Analytica chimica acta, 663(2), 165-171 (2010-03-09)
An analytical method has been developed for the simultaneous determination of the following endectocide drugs in milk: ivermectin, abamectin, doramectin, moxidectin, eprinomectin, emamectin and nemadectin. Samples were extracted with acetonitrile, purified with solid-phase extraction on a reversed phase C(8), derivatised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service