Skip to Content
Merck
All Photos(6)

Key Documents

N2127

Sigma-Aldrich

4-Nitrophenyl α-D-mannopyranoside

α-mannosidase substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

4-Nitrophenyl a-D-mannopyranoside, 4-Nitrophenyl alpha-D-mannopyranoside, pNP-alpha-D-Man, pNPalphaMan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
92210
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl α-D-mannopyranoside, α-mannosidase substrate

Assay

≥98% (TLC)

form

powder

solubility

DMF: 50 mg/mL

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2ccc(cc2)[N+]([O-])=O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11+,12+/m1/s1

InChI key

IFBHRQDFSNCLOZ-GCHJQGSQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrophenyl α-D-mannopyranoside is a chromogenic substrate for α-mannosidase.

Application

4-Nitrophenyl α-D-mannopyranoside has been used as a substrate for α-mannosidase in hydrolase activity assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Coleman et al.
Analytical biochemistry, 399(1), 7-12 (2009-12-23)
A simple and reliable continuous assay for measurement of alpha-mannosidase activity is described and demonstrated for analysis with two recombinant human enzymes using the new substrate resorufin alpha-d-mannopyranoside (Res-Man). The product of enzyme reaction, resorufin, exhibits fluorescence emission at 585
P N Kanellopoulos et al.
Journal of structural biology, 116(3), 345-355 (1996-05-01)
Concanavalin A (Con A) is the best-known plant lectin and has important in vitro biological activities arising from its specific saccharide-binding ability. Its exact biological role still remains unknown. The complexes of Con A with 4'-nitrophenyl-alpha-D-mannopyranoside (alpha-PNM) and 4'-nitrophenyl-alpha-D-glucopyranoside (alpha-PNG)
I Sielezneff et al.
Chirurgie; memoires de l'Academie de chirurgie, 124(2), 159-164 (1999-06-01)
Peritoneal colonization is a crucial event in the pathogenesis of peritonitis and its local complications. Adherence to the serosal mesothelium is mediated in a number of microorganisms derived from the digestive tract (especially E. coli) by type-1 fimbriae which have
S Howard et al.
Biochemical and biophysical research communications, 238(3), 896-898 (1997-10-23)
The stereochemical course of the hydrolyses catalysed by two alpha-mannosidases has been determined directly by 1H NMR. Synthetic substrates were incubated with the enzymes and the anomeric configuration of the initially formed product was ascertained in each case by observation
S J Hamodrakas et al.
International journal of biological macromolecules, 11(1), 17-22 (1989-02-01)
Molecular models for the complex formed between the lectin concanavalin A (Con A) and the saccharide derivative 4'-nitrophenyl-alpha-D-mannopyranoside (alpha-PNM) are presented, combining evidence from 1H-n.m.r. measurements, semi-empirical energy calculations and interactive graphics modelling. The models are in good agreement with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service