Skip to Content
Merck
All Photos(1)

Key Documents

N7006

Sigma-Aldrich

4-Nitrophenyl β-D-glucopyranoside

≥98% (TLC)

Synonym(s):

p-Nitrophenyl β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
92211
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11-,12-/m1/s1

InChI key

IFBHRQDFSNCLOZ-RMPHRYRLSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxia Wang et al.
Journal of agricultural and food chemistry, 59(20), 11219-11227 (2011-09-16)
Two novel β-glucosidases from Trichosporon asahii, named BG1 and BG2, were purified to electrophoretic homogeneity using ammonium sulfate precipitation, hydrophobic interaction, ion exchange, and gelfiltration chromatography. The molecular weight of BG1 and BG2 were estimated as 160 kDa and 30
Edy M Vilei et al.
BMC microbiology, 7, 31-31 (2007-04-19)
Contagious bovine pleuropneumonia (CBPP) caused by Mycoplasma mycoides subsp. mycoides small-colony type (SC) is among the most serious threats for livestock producers in Africa. Glycerol metabolism-associated H2O2 production seems to play a crucial role in virulence of this mycoplasma. A
Dina Jabbour et al.
Applied microbiology and biotechnology, 93(5), 1947-1956 (2011-12-08)
An open reading frame (ORF) encoding the enzyme β-glucosidase from the extremely thermophilic bacterium Fervidobacterium islandicum has been identified, cloned and sequenced. The bgl1A gene was cloned in a pET-Blue1 vector and transformed in Escherichia coli, resulting in high-level expression
Rui M F Bezerra et al.
Applied biochemistry and biotechnology, 112(3), 173-184 (2004-03-10)
The kinetics of exoglucanase (Cel7A) from Trichoderma reesei was investigated in the presence of cellobiose and 24 different enzyme/Avicel ratios for 47 h, in order to establish which of the eight available kinetic models best explained the factors involved. The
H Michlmayr et al.
Journal of applied microbiology, 108(2), 550-559 (2009-08-26)
To study glycosidase activities of a Lactobacillus brevis strain and to isolate an intracellular beta-glucosidase from this strain. Lactic acid bacteria (LAB) isolated from a commercially available starter culture preparation for malolactic fermentation were tested for beta-glycosidase activities. A strain

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service