Skip to Content
Merck
All Photos(1)

Key Documents

H2508

Sigma-Aldrich

Hippuryl-Arg

carboxypeptidase substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

N-Benzoyl-Gly-Arg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O4
CAS Number:
Molecular Weight:
335.36
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Hippuryl-Arg, carboxypeptidase substrate

Assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

NC(=N)NCCC[C@H](NC(=O)CNC(=O)c1ccccc1)C(O)=O

InChI

1S/C15H21N5O4/c16-15(17)18-8-4-7-11(14(23)24)20-12(21)9-19-13(22)10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9H2,(H,19,22)(H,20,21)(H,23,24)(H4,16,17,18)/t11-/m0/s1

InChI key

GFLCPYUSPYXNBV-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hippuryl-Arg has been used as a substrate for:
  • aminopeptidase from Trogoderma granarium
  • exopeptidase from Bactrocera oleae Gmelin, midgut of Plagiodera versicolora
  • carboxypeptidase from Pichia pastoris

Biochem/physiol Actions

Hippuryl-Arg is a synthetic substrate for carboxypeptidase B, and aminopeptidase. The enzyme carboxypeptidase acts on C terminal of arginine and lysine.

Substrates

Substrate for carboxypeptidase B and carboxypeptidase N.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Hong Liu et al.
FEBS letters, 580(22), 5137-5142 (2006-09-12)
This work reports the use of electrospray mass spectrometry for studying the conformational dynamics of enzymes by amide hydrogen/deuterium exchange (HDX) measurements. A rapid-mixing quench-flow approach allows comparisons to be made between the HDX kinetics of free enzymes with those
S Appelros et al.
Biological chemistry, 379(7), 893-898 (1998-08-15)
Recent developments in the treatment of acute pancreatitis have focused on the importance of early determination of the severity of an attack. Measuring levels of activation peptides from pancreatic proenzymes seems to be one way to predict severity. Levels of
H Schweisfurth et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 21(10), 605-609 (1983-10-01)
Kininase I (carboxypeptidase N; EC 3.4.17.3) consists of carboxypeptidase N1 (CN1) and carboxypeptidase N2 (CN2); these two enzymes can be differentiated by their activities towards hippuryl-L-arginine and hippuryl-L-lysine, respectively. A spectrophotometric assay for both carboxypeptidases in human serum is described
Expression, purification and characterization of porcine pancreatic Carboxypeptidase B from Pichia pastoris for the conversion of recombinant human insulin
Mansur M, et al.
Enzyme and Microbial Technology, 40(3), 476-480 (2007)
Proteolytic activity in Plagiodera versicolora Laicharting (Coleoptera: Chrysomelidae): Characterization of digestive proteases and effect of host plants
Zibaee A and Hajizadeh J
Journal of Asia-Pacific Entomology, 16(3), 329-334 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service