Skip to Content
Merck
All Photos(1)

Key Documents

N1514

Sigma-Aldrich

Nervonic acid

≥99% (capillary GC)

Synonym(s):

cis-15-Tetracosenoic acid, Selacholeic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)13COOH (cis)
CAS Number:
Molecular Weight:
366.62
Beilstein:
1728551
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥99% (capillary GC)

form

powder

mp

42-43 °C (lit.)

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCCCCCC(O)=O

InChI

1S/C24H46O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h9-10H,2-8,11-23H2,1H3,(H,25,26)/b10-9-

InChI key

GWHCXVQVJPWHRF-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nervonic acid has been used as a synthetic lipid supplement in vitro to study about effect of the developmental diet in Drosophila lifespan.

Biochem/physiol Actions

Nervonic acid (C24:1), a component of membrane sphingolipids and phosphatidylethanolamines, may be a useful predictor of chronic kidney disease mortality and diabetes. Nervonic acid oils are being studied for pharmaceutical, nutraceutical and industrial applications. Nervonic acid is a major component of Lunaria oil.

Packaging

Sealed ampule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Developmental diet regulates Drosophila lifespan via lipid autotoxins
Stefana MI, et al.
Nature Communications, 8(1), 1384-1384 (2017)
Y Mizushina et al.
Journal of molecular biology, 304(3), 385-395 (2000-11-25)
Unsaturated long-chain fatty acids selectively bind to the DNA binding sites of DNA polymerase beta and DNA topoisomerase II, and inhibit their activities, although the amino acid sequences of these enzymes are markedly different from each other. Computer modeling analysis
Arja T Erkkilä et al.
The Journal of craniofacial surgery, 16(5), 764-769 (2005-09-30)
The fatty acid profiles of serum lipids were examined in 53 ten-year-old cleft children. The children presented with different cleft types (cleft lip, cleft lip and palate, isolated cleft palate and submucuous cleft palate) and were recruited from the Finnish
Peroxisomal fatty acid oxidation and cellular redox.
Inderjit Singh
Methods in enzymology, 352, 361-372 (2002-07-20)
Aleix Sala-Vila et al.
The Journal of nutrition, 134(4), 868-873 (2004-03-31)
Supplementation of formulas for full-term infants with long-chain (LC) PUFA [arachidonic acid (AA) and docosahexaenoic acid (DHA)] at levels resembling human milk is recommended because they provide biochemical and functional benefits to the neonate. The objective of this work was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service