Skip to Content
Merck
All Photos(2)

Key Documents

M3525

Sigma-Aldrich

Myristoleic acid

≥99% (capillary GC)

Synonym(s):

cis-9-Tetradecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
226.36
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥99% (capillary GC)

form

liquid

refractive index

n20/D 1.4562 (lit.)

bp

144 °C/0.6 mmHg (lit.)

mp

−4.5-−4 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C14H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h5-6H,2-4,7-13H2,1H3,(H,15,16)/b6-5-

InChI key

YWWVWXASSLXJHU-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Myristoleic acid (C14:1) is a mono-unsaturated medium-chain fatty acid used as a model substrate for enzymes such as the cytochrome P450 enzyme CYP102D1; the aromatic peroxygenase from Agrocybe aegerita and oleate hydratase from Lysinibacillus fusiformis.

Packaging

Sealed ampule.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mélissa Hannauer et al.
FEBS letters, 586(1), 96-101 (2011-12-17)
Pyoverdine I (PVDI) is the major siderophore produced by Pseudomonas aeruginosa to import iron. Biosynthesis of this chelator involves non-ribosomal peptide synthetases and other enzymes. PvdQ is a periplasmic enzyme from the NTN hydrolase family and is involved in the
Bi-Na Kim et al.
Applied microbiology and biotechnology, 95(4), 929-937 (2011-12-23)
A recombinant enzyme from Lysinibacillus fusiformis was expressed, purified, and identified as an oleate hydratase because the hydration activity of the enzyme was the highest for oleic acid (with a k (cat) of 850 min(-1) and a K (m) of 540 μM)
Martin M Hanczyc et al.
Science (New York, N.Y.), 302(5645), 618-622 (2003-10-25)
The clay montmorillonite is known to catalyze the polymerization of RNA from activated ribonucleotides. Here we report that montmorillonite accelerates the spontaneous conversion of fatty acid micelles into vesicles. Clay particles often become encapsulated in these vesicles, thus providing a
Karina Krotova et al.
British journal of pharmacology, 148(5), 732-740 (2006-05-23)
1. Myristoylated pseudosubstrate of PKCzeta (mPS) - a synthetic myristoylated peptide with a sequence (13 amino acids) mimicking the endogenous PKCzeta pseudosubstrate region -- is considered a selective cell-permeable inhibitor of PKCzeta. We present strong evidence that in endothelial cells
Kwon-Young Choi et al.
The FEBS journal, 279(9), 1650-1662 (2011-12-23)
Among 33 cytochrome P450s (CYPs) of Streptomyces avermitilis, CYP102D1 encoded by the sav575 gene is naturally a unique and self-sufficient CYP. Since the native cyp102D1 gene could not be expressed well in Escherichia coli, its expression was attempted using codon-optimized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service