Skip to Content
Merck
All Photos(2)

Documents

L2376

Sigma-Aldrich

Linolenic acid

≥99%

Synonym(s):

α-Lnn, cis,cis,cis-9,12,15-Octadecatrienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7CO2H
CAS Number:
Molecular Weight:
278.43
Beilstein:
1727693
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99%

refractive index

n20/D 1.480 (lit.)

bp

230-232 °C/1 mmHg (lit.)

mp

-11 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

lipid type

omega FAs

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\CCCCCCCC(O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-

InChI key

DTOSIQBPPRVQHS-PDBXOOCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linolenic acid is an essential fatty acid, which is not produced in the body. Plants and fishes are rich sources of linolenic acid.

Application

Linolenic acid has been used:
  • to assess the total antioxidative activity of serum and bacteria
  • in palleroni chamber assay
  • to prepare free fatty acid mixture

Biochem/physiol Actions

Linolenic acid is involved in the formation of highly unsaturated fatty acid. It is essential for the development of skin, blood and neural cells. Linolenic acid is also associated with the organization of lipid membranes of neural synapses, pigment epithelial cells of retina and myocardial cells. It plays an important role in brain development and prostaglandin synthesis. Deficiency of essential fatty acid is related to skin disorders and growth retardation.
An ω-3 fatty acid that serves as a precursor to eicosapentaenoic acid (EPA) but not docosahexaenoic acid. Conversion is greater in women than men, and conversely, β-oxidation metabolism is greater in men than women.

Packaging

Sealed ampule

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Culture of rodent spermatogonial stem cells, male germline stem cells of the postnatal animal
Kubota Hiroshi and Brinster Ralph L
Methods in Cell Biology, 86, 59-84 (2008)
Rennie & Roberton's Textbook of Neonatology E-Book, 285-285 (2012)
Bacterial chemoattraction towards jasmonate plays a role in the entry of Dickeya dadantii through wounded tissues
Antunez-Lamas M, et al.
Molecular Microbiology, 74(3), 662-671 (2009)
Sabiston Textbook of Surgery E-Book, 144-144 (2012)
Two antioxidative lactobacilli strains as promising probiotics
Kullisaar T, et al.
International Journal of Food Microbiology, 72(3), 215-224 (2002)

Articles

Importance and uses of linolenic acid in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Lipid Induced Insulin Resistance

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

See All

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service