M003
R-(−)-Deprenyl hydrochloride
powder, ≥98% (HPLC)
Synonym(s):
(R)-(−)-N,α-Dimethyl-N-(2-propynyl)phenethylamine hydrochloride, R(−)-N-α-Dimethyl-N-2-propynyl-benzeneethanamine hydrochloride, Selegiline hydrochloride
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
optical activity
[α]25/D −10.8°, c = 6.48 in H2O(lit.)
color
white
mp
141-142 °C
solubility
H2O: >10 mg/mL
originator
Sanofi Aventis
SMILES string
Cl[H].C[C@H](Cc1ccccc1)N(C)CC#C
InChI
1S/C13H17N.ClH/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12H,10-11H2,2-3H3;1H/t12-;/m1./s1
InChI key
IYETZZCWLLUHIJ-UTONKHPSSA-N
Gene Information
human ... MAOB(4129)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
application for HPLCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service