Skip to Content
Merck
All Photos(4)

Key Documents

K3250

Sigma-Aldrich

Kynuramine dihydrobromide

monoamine oxidase substrate, fluorogenic, ≥97% (TLC), crystalline

Synonym(s):

3-(2-Aminophenyl)-3-oxopropanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O · 2HBr
CAS Number:
Molecular Weight:
326.03
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Kynuramine dihydrobromide, crystalline

Assay

≥97% (TLC)

form

crystalline

solubility

water: 50 mg/mL, clear to slightly hazy

storage temp.

−20°C

SMILES string

Br.NCCC(=O)c1ccccc1N

InChI

1S/C9H12N2O.BrH/c10-6-5-9(12)7-3-1-2-4-8(7)11;/h1-4H,5-6,10-11H2;1H

InChI key

YUPVVZSYBUIDQR-UHFFFAOYSA-N

Application

Kynuramine dihydrobromide has been used as a substrate for monoamine oxidase.

Biochem/physiol Actions

Kynuramine dihydrobromide is a substrate for the enzyme monoamine oxidase. The end product is the formation of fluorescence detectable 4-hydroxyquinoline (4-HOQ) measured in a fluorescence spectrophotometer at 380 nm.

Substrates

Substrate for monoamine oxidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic dietary mercury exposure causes oxidative stress, brain lesions, and altered behaviour in Atlantic salmon (Salmo salar) parr
Berntssen MHG, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 65(1), 55-72 (2003)
Magdalena S Nel et al.
Bioorganic chemistry, 69, 20-28 (2016-09-24)
In the present study a series of fifteen 2-heteroarylidene-1-indanone derivatives were synthesised and evaluated as inhibitors of recombinant human monoamine oxidase (MAO) A and B. These compounds are structurally related to series of heterocyclic chalcone derivatives which have previously been
Synergistic antidepressant-like effect of ferulic acid in combination with piperine: involvement of monoaminergic system
Li G, et al.
Metabolic Brain Disease, 30(6), 1505-1514 (2015)
Evidence for the involvement of the noradrenergic system, dopaminergic and imidazoline receptors in the antidepressant-like effect of tramadol in mice
Jesse CR, et al.
Pharmacology, Biochemistry, and Behavior, 95(3), 344-350 (2010)
Izaviany da Silva Schmitz et al.
Molecular biology reports, 46(2), 2285-2292 (2019-02-14)
Oxidative stress occurs due to an imbalance between antioxidant defenses and pro-oxidant agents in brain. This condition has been associated to the pathogenesis of several brain diseases; therefore, increasing the use of compounds that exert antioxidant activity. Thus, the objective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service