Skip to Content
Merck
All Photos(1)

Key Documents

M74253

Sigma-Aldrich

N-Methyl-N-propargylbenzylamine

97%

Synonym(s):

Pargyline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)CH2C≡CH
CAS Number:
Molecular Weight:
159.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.522 (lit.)

bp

86-88 °C/4 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CN(CC#C)Cc1ccccc1

InChI

1S/C11H13N/c1-3-9-12(2)10-11-7-5-4-6-8-11/h1,4-8H,9-10H2,2H3

InChI key

DPWPWRLQFGFJFI-UHFFFAOYSA-N

Gene Information

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergiy Konovalov et al.
Journal of ovarian research, 6(1), 75-75 (2013-10-30)
Lysine-specific demethylase 1 (LSD1, also known as KDM1A and AOF2) is a chromatin-modifying activity that catalyzes the removal of methyl groups from lysine residues in histone and non-histone proteins, regulating gene transcription. LSD1 is overexpressed in several cancer types, and
Highly stereoselective synthesis of tertiary propargylic centers and their isomerization to enantiomerically enriched allenes.
José Luis García Ruano et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(32), 9775-9779 (2012-08-01)
Masahiro Yoshida et al.
The Journal of organic chemistry, 78(4), 1687-1692 (2013-02-01)
The reaction of propargylic carbonates with 4-hydroxy-2-pyrones in the presence of a palladium catalyst is described. By the choice of the reaction temperature, two types of the substituted furo[3,2-c]pyran-4-one derivatives were regioselectively synthesized, respectively.
Francesca Reineri et al.
Journal of the American Chemical Society, 134(27), 11146-11152 (2012-06-06)
(15)N-Propargylcholine has been synthesized and hydrogenated with para-H(2). Through the application of a field cycling procedure, parahydrogen spin order is transferred to the (15)N resonance. Among the different isomers formed upon hydrogenation of (15)N-propargylcholine, only the nontransposed derivative contributes to
Gangadhara R Sareddy et al.
Oncotarget, 4(1), 18-28 (2012-12-19)
Glioma development is a multistep process, involving alterations in genetic and epigenetic mechanisms. Understanding the mechanisms and enzymes that promote epigenetic changes in gliomas are urgently needed to identify novel therapeutic targets. We examined the role of histone demethylase KDM1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service