Skip to Content
Merck
All Photos(1)

Key Documents

L9761

Sigma-Aldrich

[D-Trp6]-LH-RH

≥97% (HPLC), powder

Synonym(s):

[D-Trp6]-Luteinizing hormone releasing hormone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H82N18O13
CAS Number:
Molecular Weight:
1311.45
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@H](CO)NC(=O)[C@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](Cc6c[nH]cn6)NC(=O)[C@@H]7CCC(=O)N7)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N8CCC[C@H]8C(=O)NCC(N)=O

InChI

1S/C64H82N18O13/c1-34(2)23-46(56(88)75-45(13-7-21-69-64(66)67)63(95)82-22-8-14-52(82)62(94)72-31-53(65)85)76-58(90)48(25-36-28-70-42-11-5-3-9-40(36)42)78-57(89)47(24-35-15-17-39(84)18-16-35)77-61(93)51(32-83)81-59(91)49(26-37-29-71-43-12-6-4-10-41(37)43)79-60(92)50(27-38-30-68-33-73-38)80-55(87)44-19-20-54(86)74-44/h3-6,9-12,15-18,28-30,33-34,44-52,70-71,83-84H,7-8,13-14,19-27,31-32H2,1-2H3,(H2,65,85)(H,68,73)(H,72,94)(H,74,86)(H,75,88)(H,76,90)(H,77,93)(H,78,89)(H,79,92)(H,80,87)(H,81,91)(H4,66,67,69)/t44-,45-,46-,47-,48+,49-,50-,51-,52-/m0/s1

InChI key

VXKHXGOKWPXYNA-PGBVPBMZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Trp-Leu-Arg-Pro-Gly-NH2

Application

[D-Trp6]-LH-RH (luteinizing hormone releasing hormone) has been used to study whether the (M. – X) region in electron capture dissociation provides information on amino acid composition of polypeptides. It is also used to study the effect of LH-RH on the production of progesterone (P), estradiol (E2) or human chorionic gonadotropin (hCG) by JEG-3 choriocarcinoma cells.

Biochem/physiol Actions

Potent LH-RH agonist with enhanced biological activity due to its slower rate of degradation. Like [D-Lys6]-LH-RH, the D-Trp6 analog has been shown to be effective against cancers expressing the LH-RH receptor. However, unlike the D-Lys6 analog, it is generally used in the unconjugated form.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Arencibia et al.
International journal of oncology, 16(5), 1009-1013 (2000-04-14)
Agonistic and antagonistic analogs of luteinizing hormone-releasing hormone (LHRH) inhibit the growth of various cancers in vivo. This effect is mainly exerted through the suppression of the pituitary-gonadal axis and the creation of a state of sex steroid deprivation. In
J F Nagao et al.
Animal reproduction science, 136(1-2), 69-73 (2012-11-28)
This study aimed to determine whether deslorelin acetate could induce double ovulation in mares. In Experiment 1, eight mares were treated with prostaglandin on Day 8 (D8) after ovulation, then treated with saline or with 100 μg of a controlled-release
C Asa et al.
Reproduction in domestic animals = Zuchthygiene, 47 Suppl 6, 377-380 (2013-01-04)
North American zoos began using melengestrol acetate (MGA) implants to control reproduction in wild felids in the mid-1970s. Research linking MGA and other progestin-based contraceptives to uterine and mammary pathology in canids as well as felids resulted in a shift
Barbara Wolters et al.
Hormone research in paediatrics, 78(5-6), 304-311 (2012-12-22)
We studied the effect of gonadotropin-releasing hormone analogues (GnRHa) on weight gain as a possible side effect. We analyzed longitudinally changes in BMI-SDS in 92 children [median age 8.0 years (IQR 7.1-8.9), 88% females, mean BMI-SDS 0.69 ± 1.30] with
A von Heimendahl et al.
Reproduction in domestic animals = Zuchthygiene, 47 Suppl 6, 398-399 (2013-01-04)
The aim of this study was to evaluate the practical use of Deslorelin in a clinical setting to induce oestrus and ovulation and to assess pregnancy rates. Sixteen anoestrus bitches that were brought into the clinic were implanted subcutaneously in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service