Skip to Content
Merck
All Photos(1)

Key Documents

B3303

Sigma-Aldrich

Buserelin acetate salt

≥90% (HPLC)

Synonym(s):

des-Gly10-[D-Ser[t-Bu]6]-LH-RH ethylamide acetate salt, pGlu-His-Trp-Ser-Tyr-D-Ser(t-Bu)-Leu-Arg-Pro-NHEt acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H86N16O13 · C2H4O2
CAS Number:
Molecular Weight:
1299.48
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@H](NC(=O)[C@H]6NC(=O)CC6)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccc(cc3)O)C(=O)N[C@H](COC(C)(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N2[C@@H](CCC2)C(=O)NCC.OC(=O)C

InChI

1S/C60H86N16O13.C2H4O2/c1-7-64-57(87)48-15-11-23-76(48)58(88)41(14-10-22-65-59(61)62)69-51(81)42(24-33(2)3)70-56(86)47(31-89-60(4,5)6)75-52(82)43(25-34-16-18-37(78)19-17-34)71-55(85)46(30-77)74-53(83)44(26-35-28-66-39-13-9-8-12-38(35)39)72-54(84)45(27-36-29-63-32-67-36)73-50(80)40-20-21-49(79)68-40;1-2(3)4/h8-9,12-13,16-19,28-29,32-33,40-48,66,77-78H,7,10-11,14-15,20-27,30-31H2,1-6H3,(H,63,67)(H,64,87)(H,68,79)(H,69,81)(H,70,86)(H,71,85)(H,72,84)(H,73,80)(H,74,83)(H,75,82)(H4,61,62,65);1H3,(H,3,4)/t40-,41-,42-,43-,44-,45-,46-,47+,48-;/m0./s1

InChI key

PYMDEDHDQYLBRT-DRIHCAFSSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Ser-tBu-Leu-Arg-Pro-NHEt

Biochem/physiol Actions

Buserelin is an LH-RH agonist.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Elin Sand et al.
Cell and tissue research, 351(3), 521-534 (2012-12-21)
Gonadotropin-releasing hormone (GnRH) analogs are given to women undergoing in vitro fertilization. Case reports describing the development of chronic intestinal pseudo-obstruction and auto-antibodies against GnRH after such treatment suggest a strong association between intestinal dysfunction and GnRH analogs. No experimental
Malin Huber et al.
Fertility and sterility, 100(5), 1270-1276 (2013-08-13)
To explore the utility of using the ratio between oocyte yield and total dose of FSH, i.e., the ovarian sensitivity index (OSI), to define ovarian response patterns. Retrospective cross-sectional study. University-affiliated private center. The entire unselected cohort of 7,520 IVF/intracytoplasmic
S Zhao et al.
Gynecologic and obstetric investigation, 49(1), 52-56 (2000-01-12)
Gonadotropin-releasing hormone (GnRH) has been found to be expressed in ovaries of various species and to modulate cell differentiation in ovarian cells. GnRH agonists (GnRHa) are widely used in the stimulation protocols of assisted reproduction. In the present study, the
Tom Pickles et al.
BJU international, 110(11 Pt B), E500-E507 (2012-05-09)
What's known on the subject? and What does the study add? Previous reports, with small numbers of patients, have described the problem of incomplete testosterone suppression (>1.1 or 1.7 nmol/L) with LHRH agonists. Various predisposing factors have been suggested: different
C M Kershaw-Young et al.
Reproduction, fertility, and development, 24(8), 1093-1097 (2012-09-07)
Ovulation in camelids is induced by an unidentified protein in the seminal plasma of the male termed 'ovulation-inducing factor'. This protein has been reported to be a 14-kDa protein under reducing conditions, which, when purified from seminal plasma, induces ovulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service