Skip to Content
Merck
All Photos(1)

Key Documents

SML2856

Sigma-Aldrich

Degarelix acetate

≥98% (HPLC), powder, GnRH receptor antagonist

Synonym(s):

ASP3550 acetate, Ac-D2Nal-D4Cpa-D3Pal-Ser-4Aph(L-hydroorotyl)-D4Aph(carbamoyl)-Leu-ILys-Pro-DAla-NH2 acetate salt, FE 200486, FE200486, N-Acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-4-[[[(4S)-hexahydro-2,6-dioxo-4-pyrimidinyl]carbonyl]amino]-L-phenylalanyl-4-[(aminocarbonyl)amino]-D-phenylalanyl-L-leucyl-N6-(1-methylethyl)-L-lysyl-L-prolyl-D-alaninamide acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C82H103ClN18O16 · xC2H4O2
CAS Number:
Molecular Weight:
1632.26 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Degarelix acetate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)C[C@@H](NC(=O)[C@H](NC(=O)C)Cc7cc8c(cc7)cccc8)C(=O)N[C@H](Cc6cnccc6)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc4ccc(cc4)NC(=O)[C@H]5NC(=O)NC(=O)C5)C(=O)N[C@H](Cc3ccc(cc3)NC(=O)N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N2[C@@H](CCC2)C(=O)N[C@H](C)

InChI

1S/C82H103ClN18O16.C2H4O2/c1-45(2)35-60(72(107)92-59(16-9-10-33-87-46(3)4)80(115)101-34-12-17-68(101)79(114)88-47(5)70(84)105)93-74(109)63(38-51-23-30-58(31-24-51)91-81(85)116)95-76(111)64(39-50-21-28-57(29-22-50)90-71(106)66-42-69(104)100-82(117)99-66)97-78(113)67(44-102)98-77(112)65(41-53-13-11-32-86-43-53)96-75(110)62(37-49-19-26-56(83)27-20-49)94-73(108)61(89-48(6)103)40-52-18-25-54-14-7-8-15-55(54)36-52;1-2(3)4/h7-8,11,13-15,18-32,36,43,45-47,59-68,87,102H,9-10,12,16-17,33-35,37-42,44H2,1-6H3,(H2,84,105)(H,88,114)(H,89,103)(H,90,106)(H,92,107)(H,93,109)(H,94,108)(H,95,111)(H,96,110)(H,97,113)(H,98,112)(H3,85,91,116)(H2,99,100,104,117);1H3,(H,3,4)/t47-,59+,60+,61-,62-,63-,64+,65-,66+,67+,68+;/m1./s1

InChI key

AUTFSFUMNFDPLH-KYMMNHPFSA-N

Biochem/physiol Actions

Degarelix acetate (FE200486) is a gonadotropin-/luteinizing hormone-/follicle-stimulating hormone-releasing hormone (GnRH, LHRH, FSH-RH) receptor antagonist that inhibits plasma testosterone for more than 40 days following a single s.c. dosage of 2 mg/kg in rats and monkeys, showing longer in vivo duration than abarelix, ganirelix, cetrorelix, azaline B or D-Trp6-LHRH, and exhibiting only weak histamine-releasing potency in isolated rat mast cells. Bi-weekly s.c. dosing (2 mg/kg) is efficacious in suppressing the growth of human prostate cancer PAC120 xenograft-derived tumors in mice in vivo.
Gonadotropin-releasing hormone (GnRH, LHRH, FSH-RH) receptor antagonist with in vivo efficacy against hormone-dependent prostate cancer growth.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Broqua et al.
The Journal of pharmacology and experimental therapeutics, 301(1), 95-102 (2002-03-22)
We describe the pharmacological profile in rats and monkeys of degarelix (FE200486), a member of a new class of long-acting gonadotropin-releasing hormone (GnRH) antagonists. At single subcutaneous injections of 0.3 to 10 microg/kg in rats, degarelix produced a dose-dependent suppression
G Jiang et al.
Journal of medicinal chemistry, 44(3), 453-467 (2001-07-21)
A series of antagonists of gonadotropin-releasing hormone (GnRH) of the general formula Ac-D2Nal-D4Cpa-D3Pal-Ser-4Aph/4Amf(P)-D4Aph/D4Amf(Q)-Leu-ILys-Pro-DAla-NH2 was synthesized, characterized, and screened for duration of inhibition of luteinizing hormone release in a castrated male rat assay. Selected analogues were tested in a reporter gene
Wolfgang Koechling et al.
British journal of clinical pharmacology, 70(4), 580-587 (2010-09-16)
Early studies on gonadotrophin-releasing hormone (GnRH) antagonists pointed out histamine-mediated anaphylactic reactions as a potential adverse effect of these drug candidates. In this study we have compared the histamine-releasing potential of four approved and marketed antagonists, degarelix, cetrorelix, abarelix and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service