Skip to Content
Merck
All Photos(1)

Key Documents

L9151

Sigma-Aldrich

Lys-Lys-Lys-Lys-Lys

≥55% peptide basis, Bioreagent

Synonym(s):

Lys5, Pentalysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H62N10O6
CAS Number:
Molecular Weight:
658.88
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Lys-Lys-Lys-Lys-Lys, ≥55% peptide basis

form

powder

composition

Peptide content, ~60%

concentration

≥55% (peptide)

color

white

storage temp.

−20°C

SMILES string

NCCCCC(N)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C30H62N10O6/c31-16-6-1-11-21(36)26(41)37-22(12-2-7-17-32)27(42)38-23(13-3-8-18-33)28(43)39-24(14-4-9-19-34)29(44)40-25(30(45)46)15-5-10-20-35/h21-25H,1-20,31-36H2,(H,37,41)(H,38,42)(H,39,43)(H,40,44)(H,45,46)

InChI key

IXPHOHNWDLRFJH-UHFFFAOYSA-N

Amino Acid Sequence

Lys-Lys-Lys-Lys

Biochem/physiol Actions

Short poly-L-lysines polypeptides such as trilysine (lys3), tetralysine (tetra-L-lysine, lys4) and pentalysine (penta-L-lysine, lys5) are cationic moieties that may be used in the construction of gene delivery vectors and DNA nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiana Perocchi et al.
Nucleic acids research, 35(19), e128-e128 (2007-09-28)
Recent transcription profiling studies have revealed an unanticipatedly large proportion of antisense transcription across eukaryotic and bacterial genomes. However, the extent and significance of antisense transcripts is controversial partly because experimental artifacts are suspected. Here, we present a method to
Pentalysine-deoxyribonucleic acid interactions: a model for the general effects of ion concentrations on the interactions of proteins with nucleic acids.
T M Lohman et al.
Biochemistry, 19(15), 3522-3530 (1980-07-22)
Dan Zhao et al.
Journal of neuroinflammation, 14(1), 234-234 (2017-12-05)
It has been shown that chronic stress-induced depression is associated with exaggerated inflammatory response in the brain. Alpha7 nicotinic acetylcholine receptors (α7nAChRs) regulate the cholinergic anti-inflammatory pathway, but the role of cholinergic signaling and α7nAChR in chronic stress has not
Marisa Claudia Alvarez et al.
Clinical epigenetics, 10(1), 139-139 (2018-11-10)
In the present study, we investigated the molecular mechanisms underlying the pro-apoptotic effects of quercetin (Qu) by evaluating the effect of Qu treatment on DNA methylation and posttranslational histone modifications of genes related to the apoptosis pathway. This study was
M Roux et al.
European biophysics journal : EBJ, 16(5), 267-273 (1988-01-01)
The interaction of cationic pentalysine with phospholipid membranes was studied by using phosphorus and deuterium Nuclear Magnetic Resonance (NMR) of headgroup deuterated dimyristoyl phosphatidylcholine (DMPC) and dimyristoyl phosphatidylserine (DMPS). In the absence of pentalysine, some of the deuterium and phosphorus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service