Skip to Content
Merck
All Photos(3)

Key Documents

L5399

Sigma-Aldrich

Lipid A, diphosphoryl from Escherichia coli F583 (Rd mutant)

Synonym(s):

Diphosphoryl Lipid A from E.Coli.

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

Escherichia coli (F583 Rd mutant)

Quality Level

form

lyophilized powder

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC(=O)OC(CCCCCCCCCCC)CC(=O)OC1C(NC(=O)CC(CCCCCCCCCCC)OC(=O)CCCCCCCCCCC)C(OCC2OC(OP(O)(O)=O)C(NC(=O)CC(O)CCCCCCCCCCC)C(OC(=O)CC(O)CCCCCCCCCCC)C2O)OC(CO)C1OP(O)(O)=O

InChI

1S/C94H178N2O25P2/c1-7-13-19-25-31-37-38-44-50-56-62-68-84(103)115-78(66-60-54-48-42-35-29-23-17-11-5)72-86(105)119-92-88(96-82(101)71-77(65-59-53-47-41-34-28-22-16-10-4)114-83(102)67-61-55-49-43-36-30-24-18-12-6)93(116-79(73-97)90(92)120-122(107,108)109)113-74-80-89(106)91(118-85(104)70-76(99)64-58-52-46-40-33-27-21-15-9-3)87(94(117-80)121-123(110,111)112)95-81(100)69-75(98)63-57-51-45-39-32-26-20-14-8-2/h75-80,87-94,97-99,106H,7-74H2,1-6H3,(H,95,100)(H,96,101)(H2,107,108,109)(H2,110,111,112)

InChI key

GZQKNULLWNGMCW-UHFFFAOYSA-N

General description

Lipid A is the fundamental structure of lipopolysaccharides (LPS), present in bacteria.

Application

Lipid A, diphosphoryl from Escherichia coli F583 (Rd mutant) has been used:
  • to study the effect of various lipid A molecules on lung pericyte proliferation in vitro
  • to test on THP-1 and J774A.1 cells for neopterin and nitrite formation respectively
  • as lipopolysaccharides (LPS) derivatives in binding experiments
  • to stimulate the spleen cells and as a mitogen stimulation for toll like receptor 4 (TLR4)

Biochem/physiol Actions

Lipid A is mainly essential for the robust immunostimulatory activities of lipopolysaccharides (LPS) in human. Lipid A is the hydrophobic domain of LPS helps in anchoring LPS to the plasma membrane of bacteria. It is immunologically active and an important constituent of LPS recognised by immune system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Qureshi et al.
The Journal of biological chemistry, 258(21), 12947-12951 (1983-11-10)
Lipopolysaccharides extracted from the heptoseless mutant of Salmonella typhimurium G30/C21 were hydrolyzed with either 0.1 N HCl at 100 degrees C or treated twice with 20 mM sodium acetate, pH 4.5, at 100 degrees C for 45 min and finally
E E Ribi et al.
Journal of the National Cancer Institute, 55(5), 1253-1257 (1975-11-01)
A transplantable hepatocarcinoma of guinea pigs was used as an experimental model for immunotherapy of cancer. Earlier work showed that complete regression of 6- to 7-day-old tumors could be obtained in about 60% of cases by inoculation of the tumors
N Qureshi et al.
The Journal of biological chemistry, 257(19), 11808-11815 (1982-10-10)
Endotoxin extracted from the heptose-less mutant of Salmonella typhimurium was hydrolyzed in 0.1 N HCl in methanol/water (1:1, v/v) at 100 degrees C to yield lipid A, which was then fractionated on a Sephadex LH-20 column to yield a major
Characterization of the effects of endotoxin on macrophage tumor cell killing.
J B Weinberg et al.
Journal of immunology (Baltimore, Md. : 1950), 121(1), 72-80 (1978-07-01)
E E Ribi et al.
Cancer research, 39(11), 4756-4759 (1979-11-01)
We described elsewhere that the synergistic antitumor activity of endotoxic extracts from Re mutants of gram-negative bacteria and trehalose mycolate against guinea pig syngeneic line 10 tumor was abrogated after peptide substances accompanying these extracts had been removed. This activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service