Skip to Content
Merck
All Photos(3)

Key Documents

P7912

Sigma-Aldrich

Phloretin

≥99% (HPLC), powder, GLUT inhibitor

Synonym(s):

β-(4-Hydroxyphenyl)-2,4,6-trihydroxypropiophenone, 2′,4′,6′-Trihydroxy-3-(4-hydroxyphenyl)propiophenone, 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-1-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O5
CAS Number:
Molecular Weight:
274.27
Beilstein:
1887240
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Phloretin, ≥99%

Assay

≥99%

form

powder

mp

~260 °C

storage temp.

2-8°C

SMILES string

Oc1ccc(CCC(=O)c2c(O)cc(O)cc2O)cc1

InChI

1S/C15H14O5/c16-10-4-1-9(2-5-10)3-6-12(18)15-13(19)7-11(17)8-14(15)20/h1-2,4-5,7-8,16-17,19-20H,3,6H2

InChI key

VGEREEWJJVICBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Major polyphenol found in apple; aglycone of phloridzin.
Phloretin a polar dye belongs to the class of dihydrochalcones.

Application

Phloretin has been used:
  • to study its effect on the 2-[N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino]-2-deoxy-d-glucose (2-NBDG) and 2-[N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino]-2-deoxy-l-glucose (2-NBDLG) uptake
  • to incubate microvesicles in order to inhibit GLUT1 (glucose transporter 1)-mediated transport in radioactive ligand up-take assay
  • as a component of KRH buffer to stop glucose uptake by trophoblast cells in vitro

Biochem/physiol Actions

Blocks L-type Ca2+ channels; activates Ca2+-activated K+ channels in amphibian myelinated nerve fibers. Monocarboxylate transporter inhibitor.
Phloretin is known to decrease interfacial dipole potential of lipid bilayer. Phloretin possess antitumor action by preventing protein kinase C (PKC) activity and stimulating programmed cell death. It is found to inhibit the transport functions of GLUTs (glucose transporter) and aquaporins.
Reacts with vic-dicarbonyl compounds such as glyoxal and methylglyoxal, preventing cytotoxic conjugation with biological macromolecules.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-dependent, glucose transporter 1 mediated glucose uptake and tube formation in the human placental first trimester trophoblast cells
Basak S, et al.
Molecular and Cellular Biochemistry, 55(, pages=), 94-99 (2018)
Samer Al-Samir et al.
Journal of enzyme inhibition and medicinal chemistry, 36(1), 1602-1606 (2021-07-16)
We have studied the CO2 permeability of the erythrocyte membrane of the rat using a mass spectrometric method that employs 18 O-labelled CO2. The method yields, in addition, the intraerythrocytic carbonic anhydrase activity and the membrane HCO3- permeability. For normal rat
Uptake of a fluorescent L-glucose derivative 2-NBDLG into three-dimensionally accumulating insulinoma cells in a phloretin-sensitive manner
Sasaki A, et al.
Human Cell : Official Journal of Human Cell Research Society, 29(1), 37-45 (2016)
Modulation of lipid bilayer interfacial dipole potential by phloretin, RH421, and 6-ketocholestanol as probed by gramicidin channel conductance
Duffin RL, et al.
Langmuir, 19(4), 1439-1442 (2003)
Dan Cui et al.
ASN neuro, 12, 1759091420960550-1759091420960550 (2020-09-29)
In ischemic stroke, vasopressin hypersecretion is a critical factor of cerebral swelling and brain injury. To clarify neural mechanisms underlying ischemic stroke-evoked vasopressin hypersecretion, we observed the effect of unilateral permanent middle cerebral artery occlusion (MCAO) in rats on astrocytic

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service