Skip to Content
Merck
All Photos(4)

Documents

L8662

Sigma-Aldrich

L-Lysine monohydrochloride

98.5-101.0%, suitable for cell culture, non-animal source, meets EP, JP, USP testing specifications

Synonym(s):

(S)-2,6-Diaminohexanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
3563889
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

biological source

non-animal source

Agency

meets EP testing specifications
meets JP testing specifications
meets USP testing specifications

Assay

98.5-101.0%

form

powder

quality

meets EP, JP, USP testing specifications

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

263 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

application(s)

pharmaceutical (small molecule)

SMILES string

OC([C@@H](N)CCCCN)=O.[H]Cl

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Scientific opinion on the presence of DNA in the feed additives consisting of concentrated liquid l-lysine, l-lysine monohydrochloride and concentrated liquid l-lysine monohydrochloride produced by Escherichia coliNITE BP-02917 for all animal species (METEX NOOVISTAGO).: This study explores the presence of DNA in l-lysine monohydrochloride and its variants used in animal feed, addressing safety concerns associated with genetic material in feed products (EFSA Panel et al., 2023).

  • Safety and efficacy of a feed additive consisting of concentrated liquid l-lysine, l-lysine monohydrochloride and concentrated liquid l-lysine monohydrochloride produced by Escherichia coliNITE BP-02917 for all animal species (Metex NoovistaGo).: Evaluates the safety and efficacy of l-lysine monohydrochloride in animal feed, contributing to a better understanding of its use and regulatory compliance (EFSA Panel et al., 2022).

  • Safety and efficacy of a feed additive consisting of l-lysine monohydrochloride and l-lysine sulfate produced by fermentation with Corynebacterium glutamicumCGMCC 17927 for all animal species (Barentz Animal Nutrition B.V.).: This publication assesses the safety and effectiveness of l-lysine monohydrochloride and l-lysine sulfate in various animal feeds, ensuring they meet health and nutritional standards (EFSA Panel et al., 2022).

  • Safety and efficacy of a feed additive consisting of l-lysine monohydrochloride and l-lysine sulfate produced by Corynebacterium glutamicum CGMCC 14498 for all animal species (Kempex Holland BV).: Investigates the implications of using l-lysine monohydrochloride and l-lysine sulfate in feed, focusing on their impact on animal health and growth (EFSA Panel et al., 2021).

  • Safety and efficacy of the feed additives concentrated liquid l-lysine (base) and l-lysine monohydrochloride produced by Corynebacterium glutamicum KCCM 80183 for all animal species (CJ Europe GmbH).: Analyzes the safety and benefits of concentrated liquid l-lysine and l-lysine monohydrochloride, detailing their advantages and regulatory approval for use in feed (EFSA Panel et al., 2021).


Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guntur Fibriansah et al.
Nature communications, 6, 6341-6341 (2015-02-24)
Dengue virus (DENV) infects ~400 million people annually. There is no licensed vaccine or therapeutic drug. Only a small fraction of the total DENV-specific antibodies in a naturally occurring dengue infection consists of highly neutralizing antibodies. Here we show that
Chunaram Choudhary et al.
Nature reviews. Molecular cell biology, 15(8), 536-550 (2014-07-24)
Lysine acetylation is a conserved protein post-translational modification that links acetyl-coenzyme A metabolism and cellular signalling. Recent advances in the identification and quantification of lysine acetylation by mass spectrometry have increased our understanding of lysine acetylation, implicating it in many
Alexander Leitner et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(26), 9455-9460 (2014-06-19)
The study of proteins and protein complexes using chemical cross-linking followed by the MS identification of the cross-linked peptides has found increasingly widespread use in recent years. Thus far, such analyses have used almost exclusively homobifunctional, amine-reactive cross-linking reagents. Here
Lunzhi Dai et al.
Nature chemical biology, 10(5), 365-370 (2014-04-01)
We report the identification of a new type of histone mark, lysine 2-hydroxyisobutyrylation (Khib), and identify the mark at 63 human and mouse histone Khib sites, including 27 unique lysine sites that are not known to be modified by lysine
Bryan J Wilkins et al.
Science (New York, N.Y.), 343(6166), 77-80 (2014-01-05)
Metaphase chromosomes are visible hallmarks of mitosis, yet our understanding of their structure and of the forces shaping them is rudimentary. Phosphorylation of histone H3 serine 10 (H3 S10) by Aurora B kinase is a signature event of mitosis, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service