Skip to Content
Merck
All Photos(4)

Key Documents

608041

Sigma-Aldrich

L-Lysine-13C6,15N2 hydrochloride

99 atom % 13C, 99 atom % 15N, 95% (CP)

Synonym(s):

SILAC Amino Acid, (S)-2,6-Diaminohexanoic acid-13C6,15N2 hydrochloride, 13C and 15N Labeled Lys, 13C and 15N Labeled lysine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215N(13CH2)413CH(15NH2)13CO2H · HCl
CAS Number:
Molecular Weight:
190.59
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C
99 atom % 15N

Quality Level

Assay

95% (CP)

form

solid

optical activity

[α]25/D +20.7°, c = 2 in 5 M HCl

technique(s)

bio NMR: suitable
protein expression: suitable

mp

263-264 °C (dec.) (lit.)

mass shift

M+8

storage temp.

room temp

SMILES string

Cl.[15NH2][13CH2][13CH2][13CH2][13CH2][13C@H]([15NH2])[13C](O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1;

InChI key

BVHLGVCQOALMSV-GSMNGUNQSA-N

General description

Lysine is a basic ketogenic amino acid with a protonated alkyl amino group. Lysine degradation results in the formation of acetoacetate. Acetylation/deacetylation of lysine residues in histones is a mechanism to regulate chromatin organization in eukaryotes.[1][2] Isotope labeled amino acids are required for the production of isotopically labeled proteins.[3][4][5]

Application

  • L-Lysine-13C6,15N2 hydrochloride has been used in pSILAC (pulsed stable isotope labeling by amino acids in cell culture) mass spectrometry to study protein profile in cells transfected with microRNAs.[6]
  • It has been used in SILAC isotopic labeling for the characterization of lysine racemase.[7]
  • It has been used in SILAC labeling for phosphoproteomic measurements in neuro-2a cells treated with and without GFKP-19 (2-pyrrolidone derivative).[8]
Pulsed Stable Isotope labeling of amino acid has been used to study host-cell function, survival, the precise intracellular pathways in protein synthesis of HIV-1 infected human monocytederived macrophages.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whole cell proteome regulation by microRNAs captured in a pulsed SILAC mass spectrometry approach.
Ebner OA and Selbach M
Methods in Molecular Biology, 725, 315-331 (2011)
Deep Phosphoproteomic Measurements Pinpointing Drug Induced Protective Mechanisms in Neuronal Cells.
Yu C, et al.
Frontiers in Physiology, 7, 635-635 (2016)
Cell-specific labeling enzymes for analysis of cell-cell communication in continuous co-culture.
Tape CJ, et al.
Molecular and Cellular Proteomics, 13, 1866-1876 (2014)
Ivan V Zhukov et al.
The journal of physical chemistry letters, 11(17), 7291-7296 (2020-08-14)
Multidimensional nuclear magnetic resonance (NMR) is based on a combination of well-established building blocks for polarization transfer. These blocks are used to design correlation experiments through one or a few chemical bonds or through space. Here, we introduce a building
Alex A Makarov et al.
Nature communications, 10(1), 3056-3056 (2019-07-13)
Lamin A is a nuclear intermediate filament protein critical for nuclear architecture and mechanics and mutated in a wide range of human diseases. Yet little is known about the molecular architecture of lamins and mechanisms of their assembly. Here we

Related Content

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service