Skip to Content
Merck
All Photos(9)

Documents

HPA001122

Sigma-Aldrich

Anti-WDHD1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acidic nucleoplasmic DNA-binding protein 1 antibody produced in rabbit, Anti-And-1 antibody produced in rabbit, Anti-WD repeat and HMG-box DNA-binding protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QTLNIVTWSPCGQYLAAGSINGLIIVWNVETKDCMERVKHEKGYAICGLAWHPTCGRISYTDAEGNLGLLENVCDPSGKTSSSKVSSRVEKDYNDLFDGDDMSNAGDFLNDNAVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDHD1(11169)

General description

The gene WDHD1 (WD repeat and HMG-box DNA binding protein 1) is mapped to human chromosome 14q22. The encoded protein contains a WD40-domain and HMG-domain.

Immunogen

WD repeat and HMG-box DNA-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

WD repeat and HMG-box DNA binding protein 1 is a protein encoded by the WDHD1 gene in humans. It is a conserved protein required for DNA replication and the establishment of sister chromatid cohesion. It plays an important role in DNA replication and its ability to stimulate the replicative DNA polymerases may contribute to this effect. It also plays a crutial role in lung and esophageal carcinogenesis as a cell cycle regulator and a downstream molecule in the phosphoinositide 3-kinase/AKT pathway. WDHD1 can act as a candidate biomarker and a promising therapeutic target for cancer. It associates with centromeres at the mid-to-late S phase and regulates the functions of centromere. It is involved in the processes that facilitate centromere integrity and genomic stability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70407

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Hoon Kang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(49), 19760-19765 (2013-11-21)
Chromosome transmission fidelity 4 (Ctf4) is a conserved protein required for DNA replication. In this report, interactions between human Ctf4 (hCtf4) and the replicative helicase containing the cell division cycle 45 (Cdc45)/minichromosome maintenance 2-7 (Mcm2-7)/Go, Ichi, Nii, and San (GINS)
Thomas Rio Frio et al.
Human molecular genetics, 17(20), 3154-3165 (2008-07-22)
Dominant mutations in the gene encoding the ubiquitously-expressed splicing factor PRPF31 cause retinitis pigmentosa, a form of hereditary retinal degeneration, with reduced penetrance. We and others have previously shown that penetrance is tightly correlated with PRPF31 expression, as lymphoblastoid cell
Vladimir P Bermudez et al.
The Journal of biological chemistry, 285(13), 9493-9505 (2010-01-22)
Ctf4/AND-1 is a highly conserved gene product required for both DNA replication and the establishment of sister chromatid cohesion. In this report, we examined the mechanism of action of human Ctf4 (hCtf4) in DNA replication both in vitro and in
Nagato Sato et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(1), 226-239 (2009-12-24)
We attempted to identify novel biomarkers and therapeutic targets for lung and esophageal cancers. We screened for genes that were overexpressed in a large proportion of lung and esophageal carcinomas using a cDNA microarray representing 27,648 genes or expressed sequence
Ayse Ertay et al.
Cell death & disease, 11(11), 1001-1001 (2020-11-23)
Triple-negative breast cancer (TNBC) is the most aggressive type of breast cancer that lacks the oestrogen receptor, progesterone receptor and human epidermal growth factor receptor 2, making it difficult to target therapeutically. Targeting synthetic lethality is an alternative approach for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service