Skip to Content
Merck
All Photos(5)

Key Documents

HPA002475

Sigma-Aldrich

Anti-RPH3A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Exophilin-1 antibody produced in rabbit, Anti-Rabphilin-3A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

SPAGLRRANSVQASRPAPGSVQSPAPPQPGQPGTPGGSRPGPGPAGRFPDQKPEVAPSDPGTTAPPREERTGGVGGYPAVGAREDRMSHPSGPYSQASAAAPQPAAARQPPPPEEEEEEANSYDSDEATTLGALEFSLLYDQDNS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPH3A(22895)

General description

Rabphilin 3A homolog (RPH3A) belongs to the Rab family and GTP-binding protein (G protein) superfamily. It is present mainly in synaptic vesicle of the brain and most abundantly found in the synaptic area of the rat cerebellum, retina, and neuromuscular junction.

Immunogen

rabphilin 3A

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Rabphilin 3A homolog (RPH3A) may regulate the synaptic vesicle exocytosis and Ca2+-triggered neurotransmitter release. It is involved in the neurotransmitter release from nerve terminals in Ca2+-dependent manner. It may act with the soluble N-ethylmaleimide-sensitive-factor attachment protein receptor (SNARE) complex.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85195

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Takai et al.
Genes to cells : devoted to molecular & cellular mechanisms, 1(7), 615-632 (1996-07-01)
There exists a small GTP-binding protein (G protein) superfamily, consisting of more than 50 members, from yeast to mammal. The Rab family belongs to this superfamily and is implicated in intracellular vesicle trafficking. Rab3A small G protein is a member
Emilio González-Arnay et al.
Brain structure & function, 229(2), 359-385 (2024-01-05)
The primate hippocampus includes the dentate gyrus, cornu ammonis (CA), and subiculum. CA is subdivided into four fields (CA1-CA3, plus CA3h/hilus of the dentate gyrus) with specific pyramidal cell morphology and connections. Work in non-human mammals has shown that hippocampal
Michelle G K Tan et al.
Neurochemistry international, 64, 29-36 (2013-11-10)
Synaptic dysfunction, together with neuritic plaques, neurofibrillary tangles and cholinergic neuron loss is an established finding in the Alzheimer's disease (AD) neocortex. The synaptopathology of AD is known to involve both pre- and postsynaptic components. However, the status of rabphilin
A Mizoguchi et al.
Biochemical and biophysical research communications, 202(3), 1235-1243 (1994-08-15)
Rabphilin-3A is a putative target protein for Rab3A small GTP-binding protein which is implicated in neurotransmitter release. Rabphilin-3A is expressed mainly in brain, but its subcellular localization remains to be clarified. Immunohistochemical analysis has revealed that Rabphilin-3A is most abundant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service