Skip to Content
Merck
All Photos(1)

Key Documents

HPA016744

Sigma-Aldrich

Anti-RAPH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 9 protein, Anti-Lamellipodin, Anti-PREL-2, Anti-Proline-rich EVH1 ligand 2, Anti-Protein RMO1, Anti-RAPH1, Anti-Ras-associated and pleckstrin homology domains-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QLSDEEIDHGAEEDSDKEDQDLDKMFGAWLGELDKLTQSLDSDKPMEPVKRSPLRQETNMANFSYRFSIYNLN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAPH1(65059)

Immunogen

Ras-associated and pleckstrin homology domains-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAPH1 (Ras association (RalGDS/AF-6) and pleckstrin homology domains 1) encodes an adaptor protein termed as lamellipodin protein (Lpd) comprising of RA-PH tandem domain. Lpd is involved in the cellular motility and lamelipodial protrusion. It binds to the Ena/VASPs proteins during the assembly of actin cytoskeleton. It has been reported that Lpd may also be involved in the Ras GTPase signaling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74657

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sasagu Kurozumi et al.
Breast cancer research and treatment, 172(1), 61-68 (2018-07-30)
Ras association and pleckstrin homology domains 1 (RAPH1) is involved in cytoskeleton regulation and re-epithelialisation in invasive carcinoma and, therefore, may play a key role in carcinogenesis and metastasis. We, herein, investigated the biological and clinical significance of RAPH1 in
Yu-Chung Chang et al.
Protein & cell, 4(3), 211-219 (2013-03-14)
The adapter protein Lamellipodin (Lpd) plays an important role in cell migration. In particular, Lpd mediates lamellipodia formation by regulating actin dynamics via interacting with Ena/VASP proteins. Its RA-PH tandem domain configuration suggests that like its paralog RIAM, Lpd may
Jamie S Lin et al.
PloS one, 12(9), e0184575-e0184575 (2017-09-08)
ADP-ribosylation factor 6 (ARF6) is a small GTPase necessary for regulating cellular structure, motility, and vesicle trafficking. In several cellular systems, ARF6 was shown to regulate actin dynamics in coordination with Rac1, a Rho small GTPase. We examined the function
Meire S Batistela et al.
Molecular biology reports, 40(12), 6613-6617 (2013-09-24)
Lamellipodin protein (Lpd), encoded by the RAPH1 gene, modulates the assembly of actin cytoskeleton through its binding to the Ena/VASPs proteins, and acts in cellular motility and lamelipodial protrusion. The region where RAPH1 gene is located (2q33) is deleted in
Georgi Dimchev et al.
Journal of cell science, 133(7) (2020-02-26)
Efficient migration on adhesive surfaces involves the protrusion of lamellipodial actin networks and their subsequent stabilization by nascent adhesions. The actin-binding protein lamellipodin (Lpd) is thought to play a critical role in lamellipodium protrusion, by delivering Ena/VASP proteins onto the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service