Skip to Content
Merck
All Photos(1)

Key Documents

H7779

Sigma-Aldrich

Retinoic acid p-hydroxyanilide

≥95%

Synonym(s):

4-HPR, Fenretinide, N-(4-Hydroxyphenyl)retinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33NO2
CAS Number:
Molecular Weight:
391.55
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥95%

form

powder

color

yellow to yellow-orange

storage temp.

−20°C

SMILES string

CC1=C(\C=C\C(C)=C\C=C\C(C)=C\C(=O)Nc2ccc(O)cc2)C(C)(C)CCC1

InChI

1S/C26H33NO2/c1-19(11-16-24-21(3)10-7-17-26(24,4)5)8-6-9-20(2)18-25(29)27-22-12-14-23(28)15-13-22/h6,8-9,11-16,18,28H,7,10,17H2,1-5H3,(H,27,29)/b9-6+,16-11+,19-8+,20-18+

InChI key

AKJHMTWEGVYYSE-FXILSDISSA-N

General description

Retinoic acid p-hydroxyanilide is a synthetic analog of retinoid.

Application

Retinoic acid p-hydroxyanilide has been used:
  • as a synthetic retinoid to induce apoptosis in SEB-1 sebocytes
  • as a medium supplement for C2C12 myoblasts to test its effect on ceramide formation
  • to test in cytotoxicity in T-cell acute lymphoblastic leukemia (T-ALL)

Biochem/physiol Actions

Retinoic acid p-hydroxyanilide, also called fenretinide, increases reactive oxygen species, activates caspases and induces apoptosis. It also inhibits dihydroceramide desaturase, leading to a decrease in ceramide biosynthesis. Fenretinide may elicit anticancer activity in cultured human breast cancer cells. It acts as an insulin antagonist and may be useful in treating insulin resistance. Fenretinide or 4-HPR has chemotherapeutic potential and is cytotoxic to retinoic acid-resistant cancers.
Vitamin A acid analogue with antiproliferative activity; induces apoptosis in malignant hemopoietic cell lines.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sundy N Y Yang et al.
Antiviral research, 177, 104760-104760 (2020-03-07)
Infection by RNA viruses such as human immunodeficiency virus (HIV)-1, influenza, and dengue virus (DENV) represent a major burden for human health worldwide. Although RNA viruses replicate in the infected host cell cytoplasm, the nucleus is central to key stages
Xiaoyan Yin et al.
EBioMedicine, 51, 102520-102520 (2019-12-27)
Metabolic syndrome (MetS), the clustering of metabolic risk factors, is associated with cardiovascular disease risk. We sought to determine if dysregulation of the lipidome may contribute to metabolic risk factors. We measured 154 circulating lipid species in 658 participants from
Claude Lachance et al.
Infection and immunity, 82(5), 1778-1785 (2014-02-20)
Streptococcus suis is an important swine pathogen and an emergent zoonotic pathogen. Excessive inflammation caused by S. suis is responsible for early high mortality in septic shock-like syndrome cases. Polyunsaturated fatty acids (PUFAs) may contribute to regulating inflammatory processes. This
Chunxiao Wang et al.
Biochemical and biophysical research communications, 493(4), 1555-1559 (2017-10-11)
In the absence of approved therapeutics, Zika virus (ZIKV)'s recent prolific outbreaks in the Americas, together with impacts on unborn fetuses of infected mothers, make it a pressing human health concern worldwide. Although a key player in viral replication in
Hui Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5606-5611 (2013-03-21)
Leukemia stem cells (LSCs) play important roles in leukemia initiation, progression, and relapse, and thus represent a critical target for therapeutic intervention. However, relatively few agents have been shown to target LSCs, slowing progress in the treatment of acute myelogenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service